Skip to Content
Merck
All Photos(3)

Key Documents

M5162

Sigma-Aldrich

MOPS

BioXtra, ≥99.5% (titration)

Synonym(s):

3-(N-Morpholino)propanesulfonic acid, 4-Morpholinepropanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO4S
CAS Number:
Molecular Weight:
209.26
Beilstein:
1106776
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Assay

≥99.5% (titration)

form

powder

technique(s)

cell culture | mammalian: suitable
electrophoresis: suitable

impurities

Insoluble matter, passes filter test

ign. residue (900 °C)

≤0.1% (as SO4)

loss

≤1% loss on drying, 110°C

pH

2.5-4.0 (1 M in H2O)

useful pH range

6.5-7.9

pKa (25 °C)

7.2

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.005%

cation traces

Al: ≤0.0005%
Ba: ≤0.0005%
Bi: ≤0.0005%
Ca: ≤0.001%
Cd: ≤0.0005%
Co: ≤0.0005%
Cr: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Li: ≤0.0005%
Mg: ≤0.0005%
Mn: ≤0.0005%
Mo: ≤0.0005%
Na: ≤0.005%
Ni: ≤0.0005%
Pb: ≤0.0005%
Sr: ≤0.0005%
Zn: ≤0.0005%

absorption

≤0.015 at 280 in H2O at 1 M
≤0.020 at 260 in H2O at 1 M

λ

1 M in H2O

application(s)

diagnostic assay manufacturing

storage temp.

room temp

SMILES string

OS(=O)(=O)CCCN1CCOCC1

InChI

1S/C7H15NO4S/c9-13(10,11)7-1-2-8-3-5-12-6-4-8/h1-7H2,(H,9,10,11)

InChI key

DVLFYONBTKHTER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Micro-osteoperforations accelerate orthodontic tooth movement by stimulating periodontal ligament cell cycles.: This study explores the use of MOPS buffer in an orthodontic context to promote accelerated tooth movement, enhancing the treatment′s efficiency and patient outcomes (Sugimori et al., 2018).

  • Chitosan-Promoted Direct Electrochemistry of Human Sulfite Oxidase.: Research demonstrates MOPS buffer′s critical role in maintaining enzyme stability and activity during electrochemical studies, providing insights into biochemical applications (Kalimuthu et al., 2017).

  • Peptide fractionation by SDS-free polyacrylamide gel electrophoresis for proteomic analysis via DF-PAGE.: The study uses MOPS buffer in innovative electrophoretic techniques to improve the resolution and separation of peptides, aiding proteomic analyses (Ramos et al., 2012).

Other Notes

Easily compare specifications for MOPS products with the MOPS specification table.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y S L Lee et al.
Human reproduction (Oxford, England), 30(3), 543-552 (2015-01-09)
What is the relationship between cleavage stage embryo kinetics, blastocyst metabolism and subsequent embryo viability? Embryos cleaving faster at the first cleavage division resulted in blastocysts with a larger inner cell mass (ICM), higher glucose consumption, lower glycolytic rate, higher
Junji Suzuki et al.
Nature communications, 5, 4153-4153 (2014-06-14)
The endoplasmic reticulum (ER) and mitochondria accumulate Ca(2+) within their lumens to regulate numerous cell functions. However, determining the dynamics of intraorganellar Ca(2+) has proven to be difficult. Here we describe a family of genetically encoded Ca(2+) indicators, named calcium-measuring
Sanna Byström et al.
Journal of proteome research, 13(11), 4607-4619 (2014-09-19)
The brain is a vital organ and because it is well shielded from the outside environment, possibilities for noninvasive analysis are often limited. Instead, fluids taken from the spinal cord or circulatory system are preferred sources for the discovery of
Sewa Rijal et al.
Blood, 125(18), 2815-2824 (2015-03-05)
Phosphoinositide signaling regulates diverse cellular functions. Phosphoinositide-3 kinase (PI3K) generates PtdIns(3,4,5)P3 and PtdIns(3,4)P2, leading to the activation of proliferative and anti-apoptotic signaling pathways. Termination of phosphoinositide signaling requires hydrolysis of inositol ring phosphate groups through the actions of PtdIns(3,4,5)P3 3-phosphatase
Alexandre Albanese et al.
ACS nano, 8(6), 5515-5526 (2014-05-07)
A nanoparticle's physical and chemical properties at the time of cell contact will determine the ensuing cellular response. Aggregation and the formation of a protein corona in the extracellular environment will alter nanoparticle size, shape, and surface properties, giving it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service