Skip to Content
Merck
All Photos(7)

Key Documents

RDD001

Sigma-Aldrich

Guanidine hydrochloride

anhydrous, free-flowing, Redi-Dri, ≥99%

Synonym(s):

Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidinium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(=NH)NH2 · HCl
CAS Number:
Molecular Weight:
95.53
Beilstein:
3591990
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.28

grade

anhydrous

Quality Level

product line

Redi-Dri

Assay

≥99%

quality

free-flowing

pH

(25 °C, 4.6 - 6/573 g/L)

mp

180-185 °C (lit.)

solubility

H2O: 6 M

density

1.3 g/cm3 (lit.)

absorption

≤0.10 at 260 nm in H2O at 6 M

SMILES string

Cl[H].NC(N)=N

InChI

1S/CH5N3.ClH/c2-1(3)4;/h(H5,2,3,4);1H

InChI key

PJJJBBJSCAKJQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Strong chaotropic agent useful for the denaturation and subsequent refolding of proteins. This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yinhua Zhang et al.
BioTechniques, 70(3), 167-174 (2021-02-05)
The ongoing pandemic has demonstrated the utility of widespread surveillance and diagnostic detection of the novel SARS-CoV-2. Reverse-transcription loop-mediated isothermal amplification (RT-LAMP) has enabled broader testing, but current LAMP tests only detect single targets and require separate reactions for controls. With flu season
Trina Mouchahoir et al.
Journal of the American Society for Mass Spectrometry, 32(4), 913-928 (2021-03-13)
The Multi-Attribute Method (MAM) Consortium was initially formed as a venue to harmonize best practices, share experiences, and generate innovative methodologies to facilitate widespread integration of the MAM platform, which is an emerging ultra-high-performance liquid chromatography-mass spectrometry application. Successful implementation
Allan Joaquim Lamontanara et al.
Nature communications, 5, 5470-5470 (2014-11-18)
The activity of protein kinases is regulated by multiple molecular mechanisms, and their disruption is a common driver of oncogenesis. A central and almost universal control element of protein kinase activity is the activation loop that utilizes both conformation and
Fackson Mwale et al.
Tissue engineering. Part A, 20(21-22), 2942-2949 (2014-05-03)
Link N is a naturally occurring peptide that can stimulate proteoglycan synthesis in intervertebral disc (IVD) cells. IVD repair can also potentially be enhanced by mesenchymal stem cell (MSC) supplementation to maximize extracellular matrix (ECM) production. In a previous study
Kym Lowry et al.
PLoS pathogens, 10(6), e1004191-e1004191 (2014-06-20)
Recombination in enteroviruses provides an evolutionary mechanism for acquiring extensive regions of novel sequence, is suggested to have a role in genotype diversity and is known to have been key to the emergence of novel neuropathogenic variants of poliovirus. Despite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service