Recommended Products
species reactivity
mouse
technique(s)
ELISA: suitable
assay range
inter-assay cv: <10%
intra-assay cv: <12%
shipped in
wet ice
storage temp.
−20°C
General description
The antibody pair provided in this kit recognizes Mouse IgG4.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Annals of translational medicine, 8(21), 1369-1369 (2020-12-15)
To evaluate circulating T follicular helper (cTfh) cells and characterize their function in chronic-phase recipients after heart transplantation. Participants were divided into healthy control (HC, n=40), preoperative (Pre, n=40), and post-transplantation chronic-phase recipient (1-year, n=40) groups. The percentages of cTfh
PloS one, 14(6), e0218096-e0218096 (2019-06-18)
The interaction between atherosclerosis and commensal microbes through leaky gut syndrome (LGS), which is characterized by impaired intestinal permeability and the introduction of undesired pathogens into the body, has not been fully elucidated. Our aim was to investigate the potential
PLoS pathogens, 15(12), e1008064-e1008064 (2019-12-17)
Broadly neutralizing antibodies (bNAbs) protect against HIV infection in non-human primates and their efficacy may be enhanced through interaction with Fc receptors on immune cells. Antibody isotype is a modulator of this binding with the IgG3 subclass mediating potent Fc
Microbial pathogenesis, 129, 206-212 (2019-02-18)
Streptococcus pneumoniae infection is associated with very high morbidity and mortality throughout the world. Vaccines are an effective measure for the reduction of S. pneumoniae infection. In particular, protein vaccines are attracting increasing attention because of their good immunogenicity and
The Journal of allergy and clinical immunology, 145(6), 1574-1584 (2020-03-23)
The cause of severe nasal polyposis in aspirin-exacerbated respiratory disease (AERD) is unknown. Elevated antibody levels have been associated with disease severity in nasal polyps, but upstream drivers of local antibody production in nasal polyps are undetermined. We sought to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service