Skip to Content
Merck
All Photos(4)

Key Documents

07-463

Sigma-Aldrich

Anti-trimethyl-Histone H4 (Lys20) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H4 (trimethyl Lys20), H4K20me3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

technique(s)

ChIP: suitable
dot blot: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

trimethylation (Lys20)

Gene Information

human ... H4C1(8359)
mouse ... H4C1(326619)

General description

Histones are nuclear proteins that form octameric structures which bind DNA to form units of chromatin called nucleosomes. The family of histones—H2A, H2B, H3, and H4—are key players in gene regulation. They undergo a number of post-translational modifications (PTM) in response to various stimuli, including phosphorylation on serine and threonine residues and methylation on lysine residues. PTMs produce configural changes in histone proteins that may induce nucleosome remodeling and expose or hide DNA sequences from transcriptional complexes. Histone H4 lysine 20 (H4K20) may undergo mono-, di-, or trimethylation, which is catalyzed by the methyltransferase PR-Set7 (Set8 or KMT5a). Methylated H4K20 plays a role in regulating DNA damage responses, mitosis, DNA replication, and gene expression. Trimethylation of H4K20 contributes to gene silencing, and is a mark of the repressive heterochromatin state.

Specificity

This antibody recognizes the N-terminus of Histone H4 trimethylated at Lys20.

Immunogen

Epitope: Trimethylated Lys20
KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H4 trimethylated at Lys20.

Application

Anti-trimethyl-Histone H4 (Lys20) Antibody is a rabbit polyclonal antibody for detection of trimethyl-Histone H4 (Lys20) also known as H4K20me3, Histone H4 (tri methyl K20) & has been validated in IP, WB, ChIP, DB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Western Blot Analysis: 2 µg/mL from a representative lot detected trimethyl-Histone H4 (Lys20) in 10 µg of BALB3T3/A31 acid extract and RAT2 acid extract.

Dot Blot Specificity Analysis: Unmodified and various modified Histone peptides (see tables) were probed with Anti-trimethyl-Histone H4 (Lys20) (4 µg/mL).

Chromatin Immunoprecipitation Analysis: A representative lot immunoprecipitated Histone H4 using chromatin from HeLa cells.

Immunoprecipitation Analysis: A representative lot immunoprecipitated trimethyl-Histone H4 (Lys20) from HeLa cell lysate.

Quality

Evaluated by Western Blot in HeLa acid extract and various recombinant Histone proteins.

Western Blot Analysis: 2 µg/mL of this antibody detected trimethyl-Histone H4 (Lys20) in 10 µg of HeLa acid extract.

Target description

~13 kDa observed. An uncharacterized band may appear at ~110 kDa in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa acid extract and various recombinant Histone proteins.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service