Skip to Content
Merck
All Photos(1)

Key Documents

RAB0799

Sigma-Aldrich

Mouse IgE ELISA Kit

Synonym(s):

IgE, Immunoglobulin E

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

technique(s)

ELISA: suitable

assay range

inter-assay cv: <10%
intra-assay cv: <12%

shipped in

wet ice

storage temp.

−20°C

General description

Mouse IgE ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of a target protein in biological samples, such as serum, plasma, cell culture supernatants, urine and cell and tissue lysates. Immunoglobulin E (IgE) is involved in the acute allergic reactions. It is also involved in the chronic inflammatory allergic diseases.
The antibody pair provided in this kit recognizes Mouse IgE.

Application

Mouse IgE ELISA (Enzyme-Linked Immunosorbent Assay) Kit has been used in the quantification of immunoglobulin E (IgE) levels in bronchoalveolar lavage fluid. It has also been used in the quantification of IgE levels in the sera of mice .

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keiichiro Mine et al.
EBioMedicine, 23, 46-51 (2017-08-23)
Type 1 diabetes (T1D) is known to be caused by Th1 cell-dependent autoimmunity. Recently, we reported that TYK2 promoter variant serves as a putative virus-induced diabetes susceptibility gene associated with deteriorated interferon-dependent antiviral response. TYK2 is also related to HIES
Yinghao Niu et al.
Scientific reports, 10(1), 1962-1962 (2020-02-08)
Mechanisms mediating the protective effects of molecular hydrogen (H2) are not well understood. This study explored the possibility that H2 exerts its anti-inflammatory effect by modulating energy metabolic pathway switch. Activities of glycolytic and mitochondrial oxidative phosphorylation systems were assessed
Kenneth H Wong et al.
Immunology, 152(2), 344-355 (2017-06-06)
Blomia tropicalis is the major asthma allergen in the tropics comparable to Dermatophagoides pteronyssinus. However, little is known about the B. tropicalis epitopes recognized by T cells. Our aim was to identify the T-cell epitopes in the major B. tropicalis
Roy H E Cloots et al.
BMC pulmonary medicine, 17(1), 158-158 (2017-12-01)
(Over-)expression of arginase may limit local availability of arginine for nitric oxide synthesis. We investigated the significance of arginase1 (ARG1) for the development of airway hyperresponsiveness (AHR) and lung inflammation in female mice with ovalbumin (OVA)-induced allergic asthma. Arg1 was
Syed Nurul Hasan et al.
Science advances, 5(8), eaaw0706-eaaw0706 (2019-08-29)
Foxp3 and its protein partners establish a regulatory T (Treg) cell transcription profile and promote immunological tolerance. However, molecular features contributing to a Treg-specific gene expression program are still incompletely understood. We find that the transcription factor Bcl11b is a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service