Skip to Content
Merck
All Photos(2)

Key Documents

RAB0362

Sigma-Aldrich

Human MMP-1 ELISA Kit

for cell and tissue lysates

Synonym(s):

MMP-1, Matrix metalloproteinase-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type tissue lysate
sample type cell lysate

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 8 pg/mL
standard curve range: 24.69-18000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MMP1(4312)

General description

The Human MMP-1 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human MMP-1 cell lysate and tissue lysate.

Immunogen

Recombinant Human MMP1

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Matrix metalloproteinases (MMPs) enzymes play a central role in the maintenance and remodeling of the extracellular matrix. Elevated expression of their activity, caused either by up-regulation of their expression or down-regulation of their cognate inhibitors, has been implicated in various degenerative disorders, including arthritis, cardiovascular diseases, skeletal growth-plate disorders, and cancer metastasis. MMP-1 is a secreted collagenase with specificity toward Type I, II, III, VII, and X collagens, aggrecan, serpins, and α2-macroglobulin. It is overexpressed in lumbar intervertebral disc degeneration (IDD). It might also be associated with rheumatoid arthritis and osteoarthritis. MMP-1 is linked with juvenile idiopathic arthritis. Genetic variations in MMP-1 can affect the susceptibility and severity of this disease. MMP-1 might facilitate fibrous plaque disruption via degradation of interstitial collagens and thinning of the fibrous cap.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABDIL6ELISA 5X Sample Diluent Buffer (Item D2)SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABLYSIS12X Cell Lysis Buffer (Item J)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression profiles of MMP-1 and TIMP-1 in lumbar intervertebral disc degeneration.
Deng B
Genetics and molecular research : GMR, 14(4), 19080-19086 (2015)
Interstitial collagenase (MMP-1) expression in human carotid atherosclerosis.
Nikkari ST
Circulation, 92(6), 1393-1398 (1995)
Regulation of matrix metalloproteinase expression in tumor invasion.
Westermarck J
Faseb Journal, 13(8), 781-792 (1999)
Transcriptional regulation of collagenase (MMP-1, MMP-13) genes in arthritis: integration of complex signaling pathways for the recruitment of gene-specific transcription factors.
Vincenti MP
Arthritis Research, 4(3), 157-164 (2002)
High MMP-1, MMP-2, and MMP-9 protein levels in osteoarthritis.
Zeng GQ
Genetics and molecular research : GMR, 14(4), 14811-14822 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service