RAB0505
Human VCAM-1 ELISA Kit
for serum, plasma, cell culture supernatant and urine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type serum
sample type plasma
sample type cell culture supernatant(s)
sample type urine
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 300 pg/mL
standard curve range: 0.247-60 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... VCAM1(7412)
General description
The Human VCAM-1 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human VCAM-1 in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Human VCAM1
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Martha Lappas
Metabolism: clinical and experimental, 63(6), 860-873 (2014-04-02)
To determine the effect of maternal obesity and gestational diabetes mellitus (GDM) on the expression and release of genes involved in endothelial cell dysfunction in human placenta and omental adipose tissue. Human placenta and omental adipose tissue were obtained from
J A Zoladz et al.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society, 65(3), 441-448 (2014-06-17)
It has been demonstrated that physical training increases serum brain-derived neurotrophic factor (BDNF) in healthy people. The aim of this study was to establish the effect of physical training on the basal serum level of the BDNF in the Parkinson's
Hui Kang et al.
Bioengineered, 12(2), 10568-10579 (2021-12-04)
Rotigotine is a non-ergoline dopamine agonist that has been licensed for the treatment of Parkinson's disease. Cardiovascular diseases are the world's leading cause of death. Ox-LDL- induced endothelial damages are involved in the initiation and progression of cardiovascular diseases. In
Han Saem Jeong et al.
Phytotherapy research : PTR, 28(10), 1492-1498 (2014-04-08)
Black raspberry (Rubus occidentalis) has been known for its anti-inflammatory and anti-oxidant effects. However, short-term effects of black raspberry on lipid profiles and vascular endothelial function have not been investigated in patients with metabolic syndrome. Patients with metabolic syndrome (n = 77)
Isabella Werner et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 63(8), 649-656 (2014-05-06)
As methylene blue (MB) has been recently proposed to preserve blood pressure in case of vasoplegic syndrome and shock, an entity directly related to systemic inflammation, we aimed to elucidate the effect of MB on the expression of adhesion-molecules in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service