RAB0372
Human MMP-9 ELISA Kit
for serum, plasma, cell culture supernatant and urine
Synonym(s):
MMP-9, Matrix metalloproteinase-9
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type serum
sample type cell culture supernatant(s)
sample type plasma
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... MMP9(4318)
General description
The Human MMP-9 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human MMP-9 pro and active forms in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Human MMP9
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Biochem/physiol Actions
Matrix metalloproteinase 9 (MMP9) is a gelatinase enzyme, which catalyzes the degradation of gelatin and collagen and is preferentially inhibited by tissue inhibitor of metalloproteinases 1 (TIMP-1). The MMP9 protein is a zinc-dependent protease which is responsible for degradation of extracellular matrix components. It also participates in the pathogenesis of cancers. MMP9 is associated with various disorders, such as cystic fibrosis, acute lung injury, asthma as well as respiratory syncytial virus infection both in vitro and in vivo. MMP-9 is involved in a variety of autoimmune diseases such as systemic lupus erythematosus, rheumatoid arthritis and multiple sclerosis and be regarded as a potential therapeutic target. Mutation of MMP-9 is associated with lumbar-disc herniation (LDH) and metaphyseal anadysplasia (MAD).
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Enhanced expression of MMP-7 and MMP-9 in demyelinating multiple sclerosis lesions.
Cossins JA
Acta Neuropathologica null
Matrix Metalloproteinase-9 Mediates RSV Infection in Vitro and in Vivo.
Kong MY
Viruses null
Matrix metalloproteinases: old dogs with new tricks.
Somerville RP
Genome Biology null
Elevated circulatory MMP-2 and MMP-9 levels and activities in patients with rheumatoid arthritis and systemic lupus erythematosus.
Chang YH
Clinical Biochemistry null
Mutations in MMP9 and MMP13 determine the mode of inheritance and the clinical spectrum of metaphyseal anadysplasia.
Lausch E
American Journal of Human Genetics null
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service