Skip to Content
Merck
All Photos(2)

Key Documents

R3250

Sigma-Aldrich

Retinyl acetate

synthetic, matrix dispersion, 475,000-650,000 USP units/g

Synonym(s):

Retinol acetate, Vitamin A acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H32O2
CAS Number:
Molecular Weight:
328.49
Beilstein:
1915439
EC Number:
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058009
PubChem Substance ID:
NACRES:
NA.79
Pricing and availability is not currently available.

biological source

synthetic

form

matrix dispersion

specific activity

475,000-650,000 USP units/g

color

yellow

mp

57-58 °C

storage temp.

−20°C

SMILES string

CC1=C(/C=C/C(C)=C/C=C/C(C)=C/COC(C)=O)C(C)(C)CCC1

InChI

1S/C22H32O2/c1-17(9-7-10-18(2)14-16-24-20(4)23)12-13-21-19(3)11-8-15-22(21,5)6/h7,9-10,12-14H,8,11,15-16H2,1-6H3/b10-7+,13-12+,17-9+,18-14+

InChI key

QGNJRVVDBSJHIZ-QHLGVNSISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Retinal, retinol and retinoic acid are the aldehyde, alcohol and acid forms of vitamin A. The retinoids exist as many geometric isomers due to the unsaturated bonds in the aliphatic chain. Retinoids also exist as retinyl esters such as retinyl propionate, retinyl acetate and retinyl palmitate.[1]

Application

Retinyl acetate has been used:
  • a control diet to study its effect at different developmental periods in fish larvae[2]
  • to study its inhibitory effects on Mycobacterium avium subspecies paratuberculosis (MAP) strains[3]
  • as an internal standard in high performance liquid chromatography (HPLC) to quantify vitamin A in fortified milk[4]

Biochem/physiol Actions

Retinyl esters provide pools of vitamin A that are converted into retinol and other retinoids as required. Retinyl acetate is used in a wide range of biological applications.[1] It acts as a chemopreventive agent.[5] Retinyl acetate also has antineoplastic property.[6]

Physical form

Dispersion in cornstarch-gelatin matrix. Oil dispersible

Analysis Note

Activity based on HPLC comparison to USP standard.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vitamins A & D inhibit the growth of mycobacteria in radiometric culture
Greenstein RJ, et al.
PLoS ONE, 7(1), e29631-e29631 (2012)
A survey of vitamin A and D contents of fortified fluid milk in Ontario
Faulkner H, et al.
Journal of Dairy Science, 83(6), 1210-1216 (2000)
Intake of high levels of vitamin A and polyunsaturated fatty acids during different developmental periods modifies the expression of morphogenesis genes in European sea bass (Dicentrarchus labrax)
Villeneuve LAN, et al.
The British Journal of Nutrition, 95(4), 677-687 (2006)
Laure A N Villeneuve et al.
The British journal of nutrition, 95(4), 677-687 (2006-03-31)
The effect of the feeding period on larval development was investigated in European sea bass larvae by considering the expression level of some genes involved in morphogenesis. Larvae were fed a control diet except during three different periods (period A:
Therapeutic Use of Medicinal Plants and their Extracts:Phytochemistry and Bioactive Compounds (2018)

Questions

  1. What is the solubility information for this product? It is not dissolved in hexane, heptane, acetone, isopropyl alcohol, water, and absolute ethanol; even left overnight on stir plate.

    1 answer
    1. R3250 is classified as a matrix dispersion. This product contains gelatin, sugar, modified food starch, and other components to make it convenient as an "oil dispersible material," which will not dissolve in chloroform. The gelatin and starch components in this product would not be soluble but will be dispersed or suspended in the oil. For a soluble retinol acetate product, recommended Product No. is R4632, Retinol Acetate all trans, which is soluble in chloroform (50 mg/ml), yielding a clear, yellow-green solution. Product No. R4632 is also soluble in ethanol.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service