Skip to Content
Merck
All Photos(1)

Documents

87809

Sigma-Aldrich

Retinyl propionate

≥98.0% (sum of isomers, HPLC), ~2500 U/mg

Synonym(s):

Vitamin A propionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H34O2
CAS Number:
Molecular Weight:
342.51
Beilstein:
2475675
EC Number:
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058020
PubChem Substance ID:
NACRES:
NA.79

biological source

synthetic

Assay

≥98.0% (sum of isomers, HPLC)

form

liquid

specific activity

~2500 U/mg

contains

BHT as stabilizer

impurities

~5% cis-isomers

color

green-yellow to very dark green-yellow
yellow to very dark yellow

storage temp.

2-8°C

SMILES string

CCC(=O)OC\C=C(C)\C=C\C=C(C)\C=C\C1=C(C)CCCC1(C)C

InChI

1S/C23H34O2/c1-7-22(24)25-17-15-19(3)11-8-10-18(2)13-14-21-20(4)12-9-16-23(21,5)6/h8,10-11,13-15H,7,9,12,16-17H2,1-6H3/b11-8+,14-13+,18-10+,19-15+

InChI key

SFRPDSKECHTFQA-ONOWFSFQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Retinal, retinol and retinoic acid are the aldehyde, alcohol and acid forms of vitamin A. The retinoids exist as many geometric isomers due to the unsaturated bonds in the aliphatic chain. Retinoids also exist as retinyl esters such as retinyl propionate, retinyl acetate and retinyl palmitate.

Application

Retinyl propionate has been used to determine its effects on performance, egg quality, serum and egg oxidative status and vitamin A levels in laying quails.

Biochem/physiol Actions

Retinyl esters provide pools of vitamin A that are converted into retinol and other retinoids as needed. Retinyl propionate is used in various formulations to control wrinkles.

Unit Definition

1 international unit according to PH EUR II, 217 (1983)

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

262.4 °F - closed cup

Flash Point(C)

128 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J J J Fu et al.
The British journal of dermatology, 162(3), 647-654 (2010-04-09)
Tretinoin is considered the benchmark prescription topical therapy for improving fine facial wrinkles, but skin tolerance issues can affect patient compliance. In contrast, cosmetic antiwrinkle products are well tolerated but are generally presumed to be less efficacious than tretinoin. To
C Green et al.
Clinical and experimental dermatology, 23(4), 162-167 (1999-01-23)
The clinical and histological effects of retinyl propionate cream (a retinyl ester) on extrinsic skin ageing (photo-ageing) in man were assessed in a double-blind randomized placebo-controlled study of 80 subjects, individual parameters of this being assessed for each treatment site
S Donoghue et al.
The Journal of nutrition, 115(12), 1562-1571 (1985-12-01)
Twelve ewes were fed retinyl propionate equivalent to 12,000 micrograms retinol/kg body weight per day for the last two trimesters of pregnancy (group fed high vitamin A). Four ewes received a control level of 120 micrograms/kg body weight per day.
Retinyl propionate and climbazole combination demonstrates clinical improvement to the appearance of hyperpigmentation and deep wrinkling with minimal irritation
Hawkins S, et al.
International Journal of Cosmetic Science, 39(6), 589-599 (2017)
Effect of retinol and retinol esters on performance, egg quality, and blood and egg vitamin A levels in laying quails
Ilhan M and BULBUL T
Turkish Journal of Veterinary and Animal Sciences, 40(5), 590-597 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service