Skip to Content
Merck
All Photos(3)

Documents

R7882

Sigma-Aldrich

Retinyl acetate

BioReagent, solid or viscous liquid, synthetic, suitable for cell culture

Synonym(s):

Retinol acetate, Vitamin A acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H32O2
CAS Number:
Molecular Weight:
328.49
Beilstein:
1915439
EC Number:
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058009
PubChem Substance ID:
NACRES:
NA.75

biological source

synthetic

product line

BioReagent

Assay

≥90% (HPLC)

form

solid or viscous liquid

potency

~2,800,000 I.U. per g

mol wt

Mw 328.49 g/mol

technique(s)

cell culture | mammalian: suitable

color

light yellow to dark yellow

mp

57-58 °C

solubility

absolute ethanol: 25 mg/mL (lit.)

storage temp.

−20°C

SMILES string

CC1=C(/C=C/C(C)=C/C=C/C(C)=C/COC(C)=O)C(C)(C)CCC1

InChI

1S/C22H32O2/c1-17(9-7-10-18(2)14-16-24-20(4)23)12-13-21-19(3)11-8-15-22(21,5)6/h7,9-10,12-14H,8,11,15-16H2,1-6H3/b10-7+,13-12+,17-9+,18-14+

InChI key

QGNJRVVDBSJHIZ-QHLGVNSISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Retinyl-acetate is a natural and ester derivative of vitamin A. It is found in classical media formulations such as M-199; NCTC-109 and William′s Medium E. Vitamin A is a component of serum useful in most serum containing cell culture systems, which receive some level of vitamin A.

Application

Retinyl acetate has been used as a standard to measure the concentration of vitamin C in high performance liquid chromatography analysis (HPLC) and as a supplement in the preparation of the complete medium.

Biochem/physiol Actions

The oxidized form of retinol is retinoic acid, which has hormone-like growth factor and differentiation activity for epithelial and related cell types, especially neural cells. Retinoic acid induces embryonic stem cells to differentiate towards neural cells. Retinyl-acetate is known to be effective in treating breast cancer.

Other Notes

All trans

Preparation Note

The melting point of this material is approx. 58 °C. In order to package, it was melted and may exist as a crystalline solid or as a supercooled viscous liquid.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hormones and Vitamins in Cancer Treatment, 247-247 (1990)
Changes in the composition of yak colostrum during the first week of lactation
Mi JD, et al.
Journal of Dairy Science, 99(1), 818-824 (2016)
Using an adherent cell culture of the mouse subependymal zone to study the behavior of adult neural stem cells on a single-cell level
Ortega F, et al.
Nature Protocols, 6(12), 1847-1847 (2011)
Yong Ho Jo et al.
Asian-Australasian journal of animal sciences, 33(5), 742-752 (2020-02-15)
This study investigated the effects of vitamin A (VA) supplementation during late-stage pregnancy on longissimus dorsi muscle tissue development, birth traits, and growth performance of postnatal Korean native calves. In the preliminary experiment, twenty-six pregnant cattle (initial body weight [BW]
Natalia Cernicchiaro et al.
Foodborne pathogens and disease, 7(9), 1071-1081 (2010-05-27)
Alteration of the gastro-intestinal tract through manipulation of cattle diets has been proposed as a preharvest control measure to reduce fecal shedding of Escherichia coli O157:H7. The objective of this study was to examine the effects of the energy source's

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service