Skip to Content
Merck
All Photos(1)

Key Documents

R3155

Sigma-Aldrich

Anti-Rabbit IgG, Native−Peroxidase antibody, Mouse monoclonal

clone RabT-50, purified from hybridoma cell culture

Synonym(s):

Mouse Anti-Rabbit IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

peroxidase conjugate

antibody form

purified immunoglobulin

antibody product type

secondary antibodies

clone

RabT-50, monoclonal

form

buffered aqueous solution

species reactivity

rabbit

should not react with

human, horse, bovine, cat, rat, chicken, pig, goat, monkey

concentration

2.5-5.0 mg/mL (antibody)

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
western blot: 1:10,000-1:20,000 using 3T3 cell extracts using Anti-Actin (20-33), Catalog Number A5060, as the primary antibody
western blot: 1:10,000-1:20,000 using Hela cell extracts using Anti-Actin, N-terminal, Catalog Number A2103, as the primary antibody.

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Monoclonal Anti-Rabbit IgG, Native-Peroxidase is a purified immunoglobulin fraction of monoclonal Anti-Rabbit IgG, Native (mouse IgG1 isotype) isolated from hybridoma cell culture of the RabT-50 hybridoma, conjugated to horseradish peroxidase (HRP).
Rabbit immunoglobulins are widely used as primary antibodies in many analytical and research techniques. In immunoprecipitation assays, when target proteins are studied using rabbit primary antibody, the secondary anti-rabbit antibody used for protein detection also recognizes the heavy (50 kDa) and light (25 kDa) chains of the rabbit immunoprecipitating antibody. Consequently, this may conceal the protein of interest, as many proteins appear in the molecular weight range of the immunoglobulin heavy and light chains. The use of monoclonal anti-rabbit IgG-native-peroxidase as the secondary antibody eliminates this hindrance because the antibody is specific for non-reduced rabbit IgG and does not react with reduced rabbit IgG. Moreover, this antibody product does not react with human, monkey, bovine, horse, pig, goat, cat, rat, and chicken IgGs. Thus, this product reduces the background staining that is frequently observed with polyclonal secondary antibodies.

Specificity

Monoclonal Anti-Rabbit IgG, Native-Peroxidase reacts specifically with non-reduced rabbit IgG and does not react with reduced rabbit IgG.

Immunogen

Enriched fraction of rabbit IgG light chain.

Application

Monoclonal Anti-Rabbit IgG, Native-Peroxidase antibody produced in mouse has been used in:
  • immunoblotting
  • immunoprecipitation
  • western blotting

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 0.05% MIT.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaolin Pu et al.
Medical science monitor : international medical journal of experimental and clinical research, 26, e918382-e918382 (2020-02-01)
BACKGROUND The coatomer protein complex subunit beta 2 (COPB2) gene is upregulated and promotes cell proliferation in some cancer cells. This study aimed to investigate the role of microRNA (miRNA) targeting by COPB2 gene expression in human lung adenocarcinoma cell
A novel Cep120-dependent mechanism inhibits centriole maturation in quiescent cells
Betleja E, et al.
eLife, 7, e35439-e35439 (2018)
Yanxiang Zhang et al.
Folia neuropathologica, 58(1), 70-82 (2020-04-28)
The polarization state of microglia affects the progress of neuropathic pain. MiR-155 modulates polarization of microglia, while its role in neuropathic pain has not been well studied. We separately used lipopolysaccharide (LPS) and interleukin 4 (IL-4) for constructing an M1/M2
Deacetylation of FOXO3 by SIRT1 or SIRT2 leads to Skp2-mediated FOXO3 ubiquitination and degradation
Wang F, et al.
Oncogene, 31(12), 1546-1546 (2012)
SIRT2 suppresses adipocyte differentiation by deacetylating FOXO1 and enhancing FOXO1's repressive interaction with PPARgamma
Wang F and Tong Q
Molecular Biology of the Cell, 20(3), 801-808 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service