Skip to Content
Merck
All Photos(1)

Key Documents

A3687

Sigma-Aldrich

Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Alkaline Phosphatase Secondary Antibody, Anti Rabbit Antibody - Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, Anti Rabbit Sigma, anti rabbit antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

rabbit

technique(s)

direct ELISA: 1:30,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50
western blot: 1:30,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

IgGs are glycoprotein antibodies that modulate several immune responses. Rabbit IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-rabbit IgGs conjugated to a detectable substrate are useful tools for the analysis of target proteins. Goat anti-Rabbit IgG (whole molecule)-Alkaline Phosphatase antibody binds all rabbit Igs.

Immunogen

purified rabbit IgG as the immunogen

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Dot blot (1 paper)
Enzyme-linked immunosorbent assay (1 paper)
Immunohistochemistry (1 paper)
Western Blotting (10 papers)
Citrullination of antithrombin by PADI4 was analyzed by ELISA using alkaline phosphatase conjugated goat anti-rabbit IgG as the secondary diluted at 1:5000 in 0.05M carbonate/bicarbonate buffer (Ph 9.6).
Goat anti-Rabbit IgG (whole molecule)-Alkaline Phosphatase antibody has been used for western blot and ELISA applications.the antibody can also be used for immunohistochemistry (at 1:50, using formalin-fixed, paraffin-embedded sections).
The level of hypoxyprobe binding proteins present in retinal mouse tissue was analyzed by ELISA using alkaline phosphatase conjugated goat anti-rabbit IgG as the secondary at a dilution of 1:2000 in 5% tween/PBS. Substrate used was 4-nitrophenyl phosphate disodium salt hexahydrate (Sigma).

Physical form

Solution in 0.05 M Tris buffer, pH 8.0, containing 1 mM MgCl2, 10 mM glycine, 1% bovine serum albumin, 50% glycerol and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sônia de Fátima Soto et al.
PloS one, 12(8), e0183314-e0183314 (2017-08-19)
Female Wistar rats were exposed to filtered air (F) or to concentrated fine particulate matter (P) for 15 days. After mating, the rats were divided into four groups and again exposed to F or P (FF, FP, PF, PP) beginning
Pingxi Xiao et al.
Experimental and therapeutic medicine, 14(4), 3812-3816 (2017-10-19)
Rheumatic heart disease (RHD) occurs due to the accumulation of complications associated with rheumatic fever, and it results in high morbidity and mortality. The majority of cases of RHD are diagnosed in the chronic stages, when treatment options are limited.
Mark A Russell et al.
Islets, 5(2), 95-105 (2013-03-21)
Pro-inflammatory cytokines are important mediators of β-cell demise in type 1 diabetes, and similar mechanisms are increasingly implicated in type 2 diabetes, where a state of chronic inflammation may persist. It is likely that the actions of anti-inflammatory cytokines are
Alexander S Dowdell et al.
Journal of bacteriology, 199(6) (2017-01-11)
The Lyme disease spirochete
G Cibelli et al.
The European journal of neuroscience, 13(7), 1339-1348 (2001-04-12)
Corticotropin-releasing factor (CRF), a neuropeptide of 41 amino acids, acts as the major physiological regulator of the basal and stress-induced release of corticotropin (ACTH), beta-endorphin and other proopiomelanocortin-derived peptides from the anterior pituitary gland. In addition to its endocrine activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service