Skip to Content
Merck
All Photos(1)

Documents

P8901

Sigma-Aldrich

Phenyl-Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

form

saline suspension

Quality Level

matrix

Cross-linked 4% beaded agarose

matrix activation

epoxy

matrix attachment

amino

matrix spacer

12 atoms

capacity

20-50 mg binding capacity (human serum albumin)(per ml of packed gel)
≥4 mg binding capacity (β-lactoglobulin dimer)(per ml of packed gel)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Phenyl-agarose is used in protein chromatography, affinity chromatography and hydrophobic interactions. Phenyl-agarose has been used to further the understanding of the digestive process of the pest species Dysdercus peruvianus as well as to study early onset dementia and Alzheimer′s disease.

Physical form

Suspension in 0.5 M NaCl containing preservative

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francisco José Fernández-Gómez et al.
Journal of neuroscience research, 88(1), 155-166 (2009-07-18)
The early-onset, irreversible, severe deficits of learning and memory in the senescence-accelerated mouse (SAM)-prone/8 (SAMP8) support its use as an animal model for human dementias of early onset. Possible implication of acetylcholinesterase (AChE) and butyrylcholinesterase (BuChE) in cognitive dysfunction of
Purification and partial characterization of an aminopeptidase from the midgut tissue of Dysdercus peruvianus
Costa, I.A., et al.
Comp. Biochem. Physiol., B: Comp. Biochem., 158, 235-241 (2011)
Lukasz Bojarski et al.
Clinical chemistry and laboratory medicine, 45(10), 1273-1276 (2007-08-01)
Presenilin 1 (PS1) and presenilin 2 (PS2) are membranous proteins involved in the pathology of Alzheimer's disease. The development of specific therapies targeted at PS1 or PS2 requires the determination of biochemical properties of presenilins. Hence, in this study we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service