GE17-0965-05
Phenyl Sepharose™ 6 Fast Flow / Low Sub
Cytiva 17-0965-05, pack of 200 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106500
NACRES:
NA.56
Recommended Products
ligand
phenyl
packaging
pack of 200 mL
manufacturer/tradename
Cytiva 17-0965-05
matrix
6% cross-linked agarose
particle size
45-165 μm
average diameter
90 μm
cleaning in place
2-14
working range
3-13
suitability
suitable for bioprocess medium
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Phenyl Sepharose™ 6 Fast FLow is a well established, standard aromatic hydrophobic interaction chromatography (HIC) medium for capture and intermediate purification requiring low to medium hydrophobicity.
Application
Phenyl Sepharose™ 6 Fast FLow is part of the Sepharose™ Fast FLow HIC platform, which has been an industrial standard for HIC chromatography during recent decades.
Phenyl Sepharose™ 6 Fast FLow is composed of cross-linked 6% Agarose beads modified with standard aromatic phenyl groups via uncharged, chemically-stable ether linkages. Two levels of ligand substitution degree help to find the optimal selectivity and binding capacity for a given application.
Phenyl Sepharose™ 6 Fast FLow is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Phenyl Sepharose™ 6 Fast FLow meets industrial demands with security of supply and comprehensive technical and regulatory support.
Phenyl Sepharose™ 6 Fast FLow is composed of cross-linked 6% Agarose beads modified with standard aromatic phenyl groups via uncharged, chemically-stable ether linkages. Two levels of ligand substitution degree help to find the optimal selectivity and binding capacity for a given application.
Phenyl Sepharose™ 6 Fast FLow is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Phenyl Sepharose™ 6 Fast FLow meets industrial demands with security of supply and comprehensive technical and regulatory support.
Features and Benefits
- Standard aromatic HIC media based on Sepharose™ Fast FLow base matrix derivatized via uncharged, chemically-stable ether linkages.
- Designed for initial and intermediate purification requiring Low to medium hydrophobicity
- Two levels of substitution help find optimal selectivity and binding capacity for a given application
- BioProcess medium supported for industrial applications and well-established in approved processes
- The hydrophilic nature of the base matrix ensures Low levels of non-specific binding.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
Sepharose is a trademark of Cytiva
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service