Skip to Content
Merck
All Photos(1)

Key Documents

P8692

Sigma-Aldrich

Paromomycin sulfate salt

powder, suitable for plant cell culture, BioReagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H45N5O14 · xH2SO4
CAS Number:
Molecular Weight:
615.63 (free base basis)
Beilstein:
5715182
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

Product Name

Paromomycin sulfate salt, suitable for plant cell culture, BioReagent

product line

BioReagent

Quality Level

form

powder

technique(s)

cell culture | plant: suitable

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

agriculture

Mode of action

protein synthesis | interferes

SMILES string

O[C@H]1[C@H](O)[C@@H](CO)O[C@H](O[C@@]2([H])[C@H](O[C@@]3([H])[C@H](O)[C@H](O[C@]4([H])[C@H](N)[C@@H](O)[C@H](O)[C@H](CN)O4)[C@@H](CO)O3)[C@@H](O)[C@H](N)C[C@@H]2N)[C@@H]1N.C

InChI

1S/C23H45N5O14.CH4/c24-2-7-13(32)15(34)10(27)21(37-7)41-19-9(4-30)39-23(17(19)36)42-20-12(31)5(25)1-6(26)18(20)40-22-11(28)16(35)14(33)8(3-29)38-22;/h5-23,29-36H,1-4,24-28H2;1H4/t5-,6+,7+,8-,9-,10-,11-,12+,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23+;/m1./s1

InChI key

OYJABWUHUYVDMJ-UDXJMMFXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

Paromomycin, monomycin, is an aminoglycoside antibiotic used to select genetically transformed plants and plant cells.

Biochem/physiol Actions

Paromomycin inhibits the initiation and elongation steps of protein synthesis by binding to 16S ribosomal RNA. Paramomycin binds to the A site, which causes defective polypeptide chains to be produced and leads to cell death.

Packaging

5g,25g

Other Notes

Keep container tightly closed in a dry and well-ventilated place

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kazuki Saito et al.
Nucleic acids research, 43(9), 4591-4601 (2015-04-22)
In eukaryotes, the tRNA-mimicking polypeptide-chain release factor, eRF1, decodes stop codons on the ribosome in a complex with eRF3; this complex exhibits striking structural similarity to the tRNA-eEF1A-GTP complex. Although amino acid residues or motifs of eRF1 that are critical
Vasundhra Bhandari et al.
PLoS neglected tropical diseases, 6(5), e1657-e1657 (2012-05-26)
With widespread resistance to antimonials in Visceral Leishmaniasis (VL) in the Indian subcontinent, Miltefosine (MIL) has been introduced as the first line therapy. Surveillance of MIL susceptibility in natural populations of Leishmania donovani is vital to preserve it and support
Sarah Hendrickx et al.
PLoS neglected tropical diseases, 6(5), e1664-e1664 (2012-06-06)
Paromomycin (PMM) has recently been introduced for treatment of visceral leishmaniasis in India. Although no clinical resistance has yet been reported, proactive vigilance should be warranted. The present in vitro study compared the outcome and stability of experimental PMM-resistance induction
Takahiko Akematsu et al.
iScience, 23(1), 100749-100749 (2019-12-31)
During sexual reproduction in the ciliate, Tetrahymena thermophila, cells of complementary mating type pair ("conjugate") undergo simultaneous meiosis and fertilize each other. In both mating partners only one of the four meiotic products is "selected" to escape autophagy, and this
Ahmed Musa et al.
PLoS neglected tropical diseases, 6(6), e1674-e1674 (2012-06-23)
Alternative treatments for visceral leishmaniasis (VL) are required in East Africa. Paromomycin sulphate (PM) has been shown to be efficacious for VL treatment in India. A multi-centre randomized-controlled trial (RCT) to compare efficacy and safety of PM (20 mg/kg/day for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service