Skip to Content
Merck
All Photos(1)

Key Documents

P5057

Sigma-Aldrich

Paromomycin sulfate salt

powder, suitable for cell culture, BioReagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H45N5O14 · xH2SO4
CAS Number:
Molecular Weight:
615.63 (free base basis)
Beilstein:
5715182
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.76

product name

Paromomycin sulfate salt, powder, BioReagent, suitable for cell culture, potency: ≥675 μg per mg

product line

BioReagent

Quality Level

form

powder

potency

≥675 μg per mg

technique(s)

cell culture | mammalian: suitable

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

protein synthesis | interferes

SMILES string

O[C@H]1[C@H](O)[C@@H](CO)O[C@H](O[C@@]2([H])[C@H](O[C@@]3([H])[C@H](O)[C@H](O[C@]4([H])[C@H](N)[C@@H](O)[C@H](O)[C@H](CN)O4)[C@@H](CO)O3)[C@@H](O)[C@H](N)C[C@@H]2N)[C@@H]1N.C

InChI

1S/C23H45N5O14.CH4/c24-2-7-13(32)15(34)10(27)21(37-7)41-19-9(4-30)39-23(17(19)36)42-20-12(31)5(25)1-6(26)18(20)40-22-11(28)16(35)14(33)8(3-29)38-22;/h5-23,29-36H,1-4,24-28H2;1H4/t5-,6+,7+,8-,9-,10-,11-,12+,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23+;/m1./s1

InChI key

OYJABWUHUYVDMJ-UDXJMMFXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside
Paromomycin or aminosidine, an aminoglycoside belongs to the class of aminoglycosides. This nonabsorbable antibiotic is found at high level in the lumen of the colon.

Application

  • Paromomycin sulfate salt has been used as a:
  • reference compound in antileishmanial activity
  • RNA-binding ligand and interacts with aptamer. This interaction prevents the binding (and cutting) of dicer to RNA duplex.
Recommended for use in cell culture applications at 100mg/L.
Paromomycin sulfate salt has been used as a positive control to compare the purine analog antiviral 2′,3′-dideoxyinosine (ddI).

Biochem/physiol Actions

Paromomycin inhibits the initiation and elongation steps of protein synthesis by binding to 16S ribosomal RNA. Paramomycin binds to the A site, which causes defective polypeptide chains to be produced and leads to cell death.
Paromomycin possesses antileishmanial and antibacterial activities. It is used to cure visceral leishmaniasis (VL) and cutaneous leishmaniasis (CL). This aminocyclitol glycoside is active against gram-negative and gram-positive bacteria. It is also active against E. histolytica, D. fragilis and several cestodes, like Taenia saginata, Taenia solium etc.

Other Notes

1g,5g
Keep container tightly closed in a dry and well-ventilated place.Keep in a dry place

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antiparasitic Agents
Infectious Diseases, 2, 1345-1372 (2017)
Use of a non-adherent cell culture system for testing the effect of 2,3'-dideoxyinosine against Cryptosporidium parvum
Lawton P, et al.
FEMS Microbiology Letters, 176(1), 235-240 (1999)
Leishmaniasis
Manson's Tropical Infectious Diseases (2014)
Nanoarchitectures for Neglected Tropical Protozoal Diseases: Challenges and State of the Art
Nano- and Microscale Drug Delivery Systems, 439-480 (2017)
Paromomycin
Davidson RN, et al.
Transactions of the Royal Society of Tropical Medicine and Hygiene, 103(7), 653-660 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service