Skip to Content
Merck
All Photos(1)

Key Documents

N5764

Sigma-Aldrich

Nisin from Lactococcus lactis

potency: ≥900 IU/mg

Synonym(s):

Nisin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C143H230N42O37S7
CAS Number:
Molecular Weight:
3354.07
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

form

powder

Quality Level

potency

≥900 IU/mg

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

[H]N[C@@H]([C@@H](C)CC)C(=O)N\C(=C\C)C(=O)N[C@H]1CSC[C@H](NC(=O)[C@H](CC(C)C)NC(=O)C(=C)NC(=O)[C@@H](NC1=O)[C@@H](C)CC)C(=O)N[C@H]2[C@H](C)SC[C@H](NC(=O)CNC(=O)[C@@H]3CCCN3C2=O)C(=O)N[C@H](CCCCN)C(=O)N[C@@H]4[C@@H](C)SC[C@H](NC(=O)CNC(=O)[C@H](CCSC)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](C)NC(=O)CNC4=O)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CCSC)C(=O)N[C@@H](CCCCN)C(=O)N[C@H]5[C@H](C)SC[C@@H]6NC(=O)[C@@H](NC(=O)[C@H](C)NC5=O)[C@H](C)SC[C@H](NC(=O)[C@H](Cc7cnc[nH]7)NC6=O)C(=O)N[C@@H](CO)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](Cc8cnc[nH]8)C(=O)N[C@@H](C(C)C)C(=O)NC(=C)C(=O)N[C@@H](CCCCN)C(O)=O

InChI

1S/C143H230N42O37S7/c1-24-69(11)105(148)135(213)162-82(27-4)118(196)174-94-58-225-59-95(175-123(201)89(48-67(7)8)169-115(193)74(16)158-138(216)107(70(12)25-2)180-132(94)210)133(211)184-112-79(21)229-61-96(160-104(190)56-152-134(212)100-38-34-44-185(100)142(112)220)128(206)164-84(36-29-32-42-145)120(198)182-109-76(18)226-60-97(161-103(189)55-151-117(195)85(39-45-223-22)165-122(200)88(47-66(5)6)168-113(191)72(14)156-102(188)54-153-136(109)214)129(207)171-92(51-101(147)187)125(203)166-86(40-46-224-23)119(197)163-83(35-28-31-41-144)121(199)183-110-77(19)228-63-99-130(208)170-90(49-80-52-149-64-154-80)124(202)176-98(62-227-78(20)111(141(219)177-99)181-116(194)75(17)159-140(110)218)131(209)173-93(57-186)127(205)179-108(71(13)26-3)139(217)172-91(50-81-53-150-65-155-81)126(204)178-106(68(9)10)137(215)157-73(15)114(192)167-87(143(221)222)37-30-33-43-146/h27,52-53,64-72,75-79,83-100,105-112,186H,15-16,24-26,28-51,54-63,144-146,148H2,1-14,17-23H3,(H2,147,187)(H,149,154)(H,150,155)(H,151,195)(H,152,212)(H,153,214)(H,156,188)(H,157,215)(H,158,216)(H,159,218)(H,160,190)(H,161,189)(H,162,213)(H,163,197)(H,164,206)(H,165,200)(H,166,203)(H,167,192)(H,168,191)(H,169,193)(H,170,208)(H,171,207)(H,172,217)(H,173,209)(H,174,196)(H,175,201)(H,176,202)(H,177,219)(H,178,204)(H,179,205)(H,180,210)(H,181,194)(H,182,198)(H,183,199)(H,184,211)(H,221,222)/b82-27+/t69-,70-,71-,72-,75-,76?,77?,78?,79?,83-,84+,85-,86-,87-,88-,89-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,105-,106-,107-,108-,109+,110-,111-,112-/m0/s1

InChI key

NVNLLIYOARQCIX-ILAIHEEGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: peptide

Application

Nisin is a polycyclic lantibiotic produced by Lactococcus lactis. It is commonly used as a food preservative. It is also used as a selective agent in cell culture for the isolation of gram-negative bacteria, yeast, and molds. Nisin has been used in bioassays to study posttranslational modification.
Used to form targeted pores in cell membranes that are mediated by lipid II. Used to study the assembly and stability of nisin-lipid II pore complexes.

Biochem/physiol Actions

Nisin binds to cell wall precursor lipid components of bacteria and disrupts cell wall production. Nisin alters the cell membrane which results in the leakage of cytoplasmic components and destruction of the proton motive force.
Nisin is an antibiotic with bactericidal action. It binds to the lipid A portion of bacterial lipopolysaccharides and induces pore formation in the membranes of cortex cells from excised sorghum roots.

Mode of Action: Binds to and interferes with the permeability of the cytoplasmic membrane.
Antimicrobial spectrum: Gram-negative and Gram-positive bacteria (requires chelator such as EDTA).

Packaging

Package size based on weight of total solids

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olli Koponen et al.
Microbiology (Reading, England), 148(Pt 11), 3561-3568 (2002-11-13)
Nisin produced by Lactococcus lactis subsp. lactis is a 34-residue antibacterial polypeptide and belongs to a group of post-translationally modified peptides, lantibiotics, with dehydrated residues and cyclic amino acids, lanthionines. These modifications are supposed to be made by enzymes encoded
Qi Zhang et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(45), 18361-18366 (2012-10-17)
Lanthionine-containing peptides (lanthipeptides) are a family of ribosomally synthesized and posttranslationally modified peptides containing (methyl)lanthionine residues. Here we present a phylogenomic study of the four currently known classes of lanthipeptide synthetases (LanB and LanC for class I, LanM for class
Michaela Wenzel et al.
Antimicrobial agents and chemotherapy, 56(11), 5749-5757 (2012-08-29)
Mersacidin, gallidermin, and nisin are lantibiotics, antimicrobial peptides containing lanthionine. They show potent antibacterial activity. All three interfere with cell wall biosynthesis by binding lipid II, but they display different levels of interaction with the cytoplasmic membrane. On one end
Des Field et al.
PloS one, 7(10), e46884-e46884 (2012-10-12)
Nisin is a bacteriocin widely utilized in more than 50 countries as a safe and natural antibacterial food preservative. It is the most extensively studied bacteriocin, having undergone decades of bioengineering with a view to improving function and physicochemical properties.
Paul D Cotter
Bioengineered, 3(6), 313-319 (2012-08-28)
While the bacteriocin Nisin has been employed by the food industry for 60 y, it remains the only bacteriocin to be extensively employed as a food preservative. This is despite the fact that the activity of Nisin against several food

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service