Skip to Content
Merck
All Photos(3)

Key Documents

P7412

Sigma-Aldrich

Pirenzepine dihydrochloride

≥98% (TLC), powder

Synonym(s):

5,11-Dihydro-11-[(4-methyl-1-piperazinyl)acetyl]-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21N5O2 · 2HCl
CAS Number:
Molecular Weight:
424.32
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

powder

color

white to light yellow

solubility

H2O: 50 mg/mL

SMILES string

Cl[H].Cl[H].CN1CCN(CC1)CC(=O)N2c3ccccc3C(=O)Nc4cccnc24

InChI

1S/C19H21N5O2.2ClH/c1-22-9-11-23(12-10-22)13-17(25)24-16-7-3-2-5-14(16)19(26)21-15-6-4-8-20-18(15)24;;/h2-8H,9-13H2,1H3,(H,21,26);2*1H

InChI key

FFNMBRCFFADNAO-UHFFFAOYSA-N

Gene Information

human ... CHRM1(1128)

Related Categories

General description

Pirenzepine dihydrochloride is an anticholinergic agent. It is also considered as an antiulcer drug. It is used to treat myopia, gastric and duodenal ulcers. Pirenzepine dihydrochloride induces the dimerization of muscarinic M1 receptors.

Application

Pirenzepine dihydrochloride has been used
  • in transactivation of fibroblast growth factor receptor (FGFR).
  • as a supplement in cell culture.
  • for the stimulation of human fibroblasts

Biochem/physiol Actions

Selective M1 muscarinic acetylcholine receptor antagonist.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Acetylcholine Receptors (Muscarinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

Hygroscopic; store desiccated at room temperature.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Corinne G Jolivalt et al.
The Journal of pharmacology and experimental therapeutics, 374(1), 44-51 (2020-04-25)
Muscarinic antagonists promote sensory neurite outgrowth in vitro and prevent and/or reverse multiple indices of peripheral neuropathy in rodent models of diabetes, chemotherapy-induced peripheral neuropathy, and HIV protein-induced neuropathy when delivered systemically. We measured plasma concentrations of the M1 receptor-selective
A Elhusseiny et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 19(7), 794-802 (1999-07-21)
Acetylcholine is an important regulator of local cerebral blood flow. There is, however, limited information available on the possible sites of action of this neurotransmitter on brain intraparenchymal microvessels. In this study, a combination of molecular and functional approaches was
Pirenzepine
Carmine AA and Brogden RN
Drugs, 30(2), 85-126 (1985)
Muscarinic acetylcholine type 1 receptor activity constrains neurite outgrowth by inhibiting microtubule polymerization and mitochondrial trafficking in adult sensory neurons
Sabbir MG, et al.
Frontiers in Neuroscience, 12(3), 229-237 (2018)
Pirenzepine promotes the dimerization of muscarinic M1 receptors through a three-step binding process
Ilien B, et al.
The Journal of Biological Chemistry, 13(6), jbc-M109 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service