Skip to Content
Merck
All Photos(1)

Key Documents

P5208

Sigma-Aldrich

Platelet-Derived Endothelial Cell Growth Factor human

≥97% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture

Synonym(s):

PD-ECGF

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Platelet-Derived Endothelial Cell Growth Factor human, recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

5-30 ng/mL EC50

mol wt

49 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

<1 EU/mg

UniProt accession no.

storage temp.

−20°C

Gene Information

General description

Platelet-Derived Endothelial Cell Growth Factor (PD-ECGF) is an endothelial cell mitogen which was originally purified from human platelets. In contrast with the endothelial mitogens of the fibroblast growth factor (FGF) family, PD-ECGF is an endothelial cell growth factor initially purified from human platelets, which does not bind to heparin and does not stimulate the proliferation of fibroblasts. This is in contrast with the effect of PDGF, which stimulates the growth of human foreskin fibroblasts, but is inactive on endothelial cells. PD-ECGF will stimulate endothelial cells in vitro and in vivo. PD-ECGF is chemotactic for bovine aortic endothelial cells. PD-ECGF does not induce smooth muscle cell migration. PD-ECGF is involved in angiogenesis and has potent angiogenic activity both in the developing vascular system of the chick chorioallantoic membrane and in vascularization of tumors in nude mice. PD-ECGF has a pI of 4.0-4.8 and a molecular weight of 49 kDa.

The biological activity of recombinant, human PD-ECGF is measured in a cell proliferation assay using human umbilical vein endothelial cells. The EC50 is defined as the effective concentration of growth factor that elicits a 50% increase in cell growth in a cell based bioassay

Application

PD-ECGF is involved in angiogenesis and has potent angiogenic activity both in the developing vascular system of the chick chorioallantoic membrane and in vascularization of tumors in nude mice.

Biochem/physiol Actions

Platelet-derived endothelial cell growth factor is a 45 kDa endothelial cell mitogen of pI 4.0-4.8 originally purified from human platelets. PD-ECGF is chemotactic for bovine aortic endothelial cells, however it does not induce smooth muscle cell migration. PD-ECGF is involved in angiogenesis and has potent angiogenic activity both in the developing vascular system of the chick chorioalantoic membrane and vascularization of tumor cells in nude mice. PD-ECGF is highly expressed in some tumors.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 0.5 mg bovine serum albumin.

Analysis Note

The proliferative activity of human PD-ECGF is measured in culture by using human umbilical vein endothelial cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Griffiths et al.
British journal of cancer, 76(6), 689-693 (1997-01-01)
Angiogenesis plays an important role in the growth and metastasis of solid tumours. Platelet-derived endothelial cell growth factor (PD-ECGF) is known to be chemotactic for endothelial cells in vitro and angiogenic in vivo. It is also known as gliostatin, a
Jue Li et al.
Bioengineered, 13(3), 4844-4856 (2022-02-15)
The association between spinal cord injury (SCI) and bladder symptoms has been intensively described. Human umbilical cord mesenchymal stem cell (hUC-MSC) treatment is beneficial to the recovery of bladder function after SCI, but its mechanism is unclear. We established an
H Aikawa et al.
Anticancer research, 19(5B), 4305-4309 (2000-01-11)
In order to elucidate the roles of tumor angiogenesis in lung carcinogenesis, the expressions of several angiogenic factors in lung carcinoma tissues were examined. Tissue specimens from 112 cases of resected non-small cell lung cancer (NSCLC) were studied. The expressions
C-S Bao et al.
European review for medical and pharmacological sciences, 22(19), 6436-6447 (2018-10-20)
This research aimed to investigate the therapeutic effects of transplanted human umbilical cord mesenchymal stem cells (hUCMSCs) on spinal cord injury in mice and to explore its molecular mechanism. Spinal cord injury model in C57BL/6J mice was established. On the
Sinsuk Han et al.
Autophagy, 17(12), 4182-4201 (2021-03-25)
Mitochondria are the main cellular energy powerhouses and supply most of the energy in the form of ATP to fuel essential neuronal functions through oxidative phosphorylation (OXPHOS). In Alzheimer disease (AD), metabolic and mitochondrial disruptions are an early feature preceding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service