Skip to Content
Merck
All Photos(1)

Key Documents

P3076

Sigma-Aldrich

Platelet-Derived Growth Factor-AA human

≥98% (SDS-PAGE and HPLC), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

PDGF−AA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

product name

Platelet-Derived Growth Factor-AA human, PDGF-AA, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE and HPLC)

form

lyophilized powder

potency

1.0-3.0 ng/mL ED50

quality

endotoxin tested

mol wt

28.5 kDa (containing 250 amino acid residues)

packaging

pkg of 10 μg
pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

<1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... PDGFRA(5154)

Looking for similar products? Visit Product Comparison Guide

General description

Platelet-derived growth factor (PDGF) is composed two polypeptide subunits of A and B chain. PDGF is produced by a number of cells including smooth muscle cells, endothelial cells and macrophages. The PDGF gene is highly conserved and is mapped to human chromosome 7p22.3. PDGF is structurally and functionally related to vascular endothelial growth factors.

Biochem/physiol Actions

Cell division and relocalization of pulmonary artery smooth muscle cells and fibroblasts is known to be triggered by platelet-derived growth factor (PDGF). Due to its proliferatory function, it is associated with fibroproliferative disorders such as atherosclerosis, pulmonary arterial hypertension and lung fibrosis. PDGF exists both in homo and heterodimeric forms and stimulates α and β-cell surface receptors. Vascular endothelial growth factor expression is regulated by PDGF-AA autocrine signalling and is known to promote tumor angiogenesis in vivo.

Other Notes

Homodimer of the A chain of human PDGF.

Physical form

Sterile-filtered through a 0.2 μm filter. Lyophilized with 10 mM acetic acid.

Analysis Note

The biological activity is measured by the dose-dependent stimulation of thymidine uptake by BALB/c 3T3 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Role of platelet-derived growth factors in physiology and medicine.
Andrae J, et al.
Genes & Development, 22.10, 1276-1312 (2008)
Clinical Findings Associated with a De Novo Partial Trisomy 10p11. 22p15. 3 and Monosomy 7p22. 3 Detected by Chromosomal Microarray Analysis.
Kohannim O, et al.
Case Reports in Genetics, 2011 (2011)
Platelet-derived growth factor-AA is an essential and autocrine regulator of vascular endothelial growth factor expression in non?small cell lung carcinomas.
Shikada Y, et al.
Cancer Research, 65.16, 7241-7248 (2005)
Luana Abballe et al.
Frontiers in pediatrics, 6, 315-315 (2018-11-18)
Numb is an intracellular protein with multiple functions. The two prevalent isoforms, Numb p66 and Numb p72, are regulators of differentiation and proliferation in neuronal development. Additionally, Numb functions as cell fate determinant of stem cells and cancer stem cells
Neha Garg et al.
The EMBO journal, 32(21), 2819-2832 (2013-10-01)
The transcription factor Nanog plays a critical role in the self-renewal of embryonic stem cells as well as in neural stem cells (NSCs). microRNAs (miRNAs) are also involved in stemness regulation. However, the miRNA network downstream of Nanog is still

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service