Skip to Content
Merck
All Photos(6)

Documents

P0899

Sigma-Aldrich

Poly-D-lysine hydrobromide

suitable for cell culture, Mol wt 70,000-150,000

Synonym(s):

D-Lysine homopolymer hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
D-Lys-(D-Lys)n-D-Lys · xHBr
CAS Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Poly-D-lysine hydrobromide, mol wt 70,000-150,000

form

solid

mol wt

70,000-150,000

technique(s)

cell culture | mammalian: suitable

impurities

<10% water (Karl Fischer)

color

white to off-white

solubility

water: 50 mg/mL, clear, colorless to very faintly yellow

functional group

amide

storage temp.

−20°C

SMILES string

O=C(C)[C@@](NC)([H])CCCCN.[Br]

InChI

1S/C6H14N2O2.BrH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H

InChI key

MEXAGTSTSPYCEP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly-D-lysine hydrobromide (PDLHB) is a water-soluble, positively charged polyamino acid that has a variety of applications in biology and biomedicine. PDLHB is commonly used to promote cell adhesion to surfaces, which is essential for cell culture, microscopy, and neuroscience research. PDLHB is also used in tissue engineering and drug delivery research. Poly-D-lysine′s primary function lies in facilitating robust cell adhesion to surfaces, leveraging its positive charge to attract and anchor cells by binding with negatively charged proteins and molecules on cell surfaces. This feature is especially valuable when coating cell culture dishes and plates, where it proves essential for ensuring reliable cell adhesion, especially for challenging cell types like primary cells and specific cancer cells. In microscopy and neuroscience applications, PDLHB-coated substrates come to the forefront, allowing for the seamless attachment of cells, tissues, and neurons to slides and culture dishes. This fosters meticulous observation and exploration of these crucial biological components, making PDLHB an indispensable resource for in-depth investigations. Furthermore, PDLHB extends its utility to tissue engineering by supporting the creation of cell growth scaffolds. These PDLHB-coated scaffolds closely replicate the natural cellular environment, promoting effective healing and regeneration. In the realm of drug delivery, PDLHB plays a pivotal role, acting as a protective shield for encapsulated drugs and precisely guiding them to specific cells and tissues. This versatile and biocompatible polymer is an invaluable asset, elevating the standards of cell culture and biochemical research+R8 within the field of biology and biomedicine.

Application

Poly-D-lysine hydrobromide is used for the following applications:
  • Used in animal cell culture
  • Rat cortical neuron cultures
  • Cell culture and siRNA treatment
  • Primary cell cultures and adult neuronal network model
  • Cortical astrocyte culture
  • Immunofluorescence staining (HDF cells were plated onto poly-D lysine (P0899, Sigma-Aldrich) coated glass coverslips)
  • NPC collection and culture conditions
Poly-D-lysine polymers can be used in preparing surfaces for cell attachment. The D-lysine polymers can also be used with cells that digest poly-L-lysine polymers and cause an excessive uptake of L-lysine.

This product is recommended as a cell culture substratum when using 0.5 - 1.0 mL of a 0.1 mg/mL solution to coat 25 cm2. Lower molecular weight versions of the product are less viscous, but high more molecular weight versions provide more attachment sites per molecule.

Biochem/physiol Actions

Poly-D-lysine (PDL) hydrobromide is a nonspecific attachment factor for cells useful in promoting cell adhesion to solid substrates by enhancing electrostatic interaction between negatively charged ions of the cell membrane and the culture surface. After absorption to the culture surface, poly-D-lysine increases the number of positively charged cell binding sites.

Features and Benefits

  • Suitable for Cell Culture and Biochemical research
  • High-quality compound suitable for multiple research applications

Components

Poly-D-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue. The hydrobromide allows the poly-D-lysine to be in a crystalline form soluble in water. A small amount of product may be found in the ß structure because the HBr interferes with hydrogen bonding between amino and either the carboxyl groups or N or O containing moieties.

Caution

Sterile solutions are stable for up to 2 years when stored at 2-8°C. It should be stored desiccated at -20°C.

Analysis Note

This product has a molecular weight of 70,000-150,000. To remove the HBr, dissolve this product in a neutral buffer and dialyze to remove the salts. In general, to use this product as an attachment factor, add 50 mL of sterile tissue culture grade water to 5 mg of poly-lysine, and aseptically coat the surface with 1 mL per 25 cm2 of solution. After 5 minutes, remove the solution through aspiration and thoroughly rinse the surface. Let dry for two hours before introducing cells and medium.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Faith C Blum et al.
Infection and immunity, 80(5), 1662-1669 (2012-03-07)
Botulinum neurotoxins (BoNTs) and tetanus neurotoxin (TeNT) are the most toxic proteins for humans. While BoNTs cause flaccid paralysis, TeNT causes spastic paralysis. Characterized BoNT serotypes enter neurons upon binding dual receptors, a ganglioside and a neuron-specific protein, either synaptic
Hyun-Joo Seo et al.
BMB reports, 41(7), 523-528 (2008-08-07)
BMI-1026 is a synthetic aminopyrimidine compound that targets cyclin dependent kinases (cdks) and was initially designed as a potential anticancer drug. Even though it has been well documented that BMI-1026 is a potent cdk inhibitor, little is known about the
Na Zhang et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 6(9), 1800808-1800808 (2019-05-09)
MicroRNAs effectively modulate protein expression and cellular response. Unfortunately, the lack of robust nonviral delivery platforms has limited the therapeutic application of microRNAs. Additionally, there is a shortage of drug-screening platforms that are directly translatable from in vitro to in
Laura Kerosuo et al.
Journal of cell science, 121(Pt 23), 3941-3950 (2008-11-13)
The mechanisms underlying the decision of a stem or progenitor cell to either self-renew or differentiate are incompletely understood. To address the role of Myc in this process, we expressed different forms of the proto-oncogene Myc in multipotent neural progenitor
Giuseppe Pani et al.
PloS one, 8(9), e73857-e73857 (2013-09-26)
One of the objectives of the current international space programmes is to investigate the possible effects of the space environment on the crew health. The aim of this work was to assess the particular effects of simulated microgravity on mature

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service