Skip to Content
Merck

A-005-M

Poly-ʟ-Lysine Hydrobromide

synthetic, liquid, 0.1 mg/mL, suitable for cell culture

Synonym(s):

Lysine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352209
eCl@ss:
32160801
NACRES:
NA.75
Biological source:
synthetic
Form:
liquid
Technique(s):
cell attachment: suitable, cell culture | mammalian: suitable
Concentration:
0.01 % (w/v), 0.1 mg/mL
Mol wt:
Mw 84000 Da
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Poly-L-Lysine Solution, 0.01%

biological source

synthetic

Quality Segment

needle size

2.0 mm

sterility

sterile

form

liquid

mol wt

Mw 84000 Da

manufacturer/tradename

NovaSeptum®

concentration

0.01 % (w/v), 0.1 mg/mL

technique(s)

cell attachment: suitable, cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Poly-L-Lysine is a synthetic amino acid chain that is positively charged having one hydrobromide per unit of Lysine. Poly-L-Lysine is widely used as a coating to enhance cell attachment and adhesion to both plasticware and glass surfaces. Certain cell types secrete proteases, which can digest Poly-L-Lysine.
The molecular weight of Poly-L-Lysine for cell culture can vary significantly, with lower molecular weight (30,000 Da) being less viscous and higher molecular weight (greater than 300,000 Da) having more binding sites per molecule. This product uses a Poly-L-Lysine of 84000 Da, yielding a solution viscosity for easy handling while providing sufficient binding sites for cell attachment.

Application

Poly-L-Lysine Solution has been used to coat culture dishes to culture U2-OS cells and primary cortical neurons.

Biochem/physiol Actions

Poly-L-lysine serves as a coating substance to ensure better attachment of cells to culture surfaces. Research shows that plates coated with poly-L-lysine (PLL) demonstrated an accelerated cell growth rate. Culturing mesenchymal stem cells (MSCs) on PLL-coated substrates has been found to safeguard the stemness properties of these cells and inhibit senescence. PLL-coated plates have demonstrated enhanced MSC growth and up-regulated the expression of genes associated with cell adhesion, differentiation, proliferation, and signaling. Moreover, porous PLL-coated surfaces have shown improvements in cell proliferation and the osteogenic differentiation of MSCs.

Preparation Note

Optimal coating concentrations must be determined by the end user. Typical coating concentrations range from 10ug/ml to 100ug/ml based on cell type and application.

1. Thaw Poly-L-Lysine solution at room temperature.

2. Dilute Poly-L-Lysine solution to the desired concentration in sterile water.

3. Fully coat the cell culture surface with diluted Poly-L-Lysine solution. Use 5 mL volume for 6-cm plates and 10 mL volume for 10-cm plates and T75 flasks.

4. Allow the cell culture vessel to sit at room temperature overnight.

5. Aspirate the Poly-L-Lysine solution the following day and rinse the vessel with sterile water followed by coating with desired ECM protein.

Analysis Note

Solution is clear with no particulates present. Solution is sterile and suitable for cell culture applications.

Legal Information

NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Poly-L-Lysine Solution


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library