Skip to Content
Merck
All Photos(5)

Key Documents

P0296

Sigma-Aldrich

Poly-D-lysine hydrobromide

suitable for cell culture, Mol wt 1,000-5,000

Synonym(s):

D-Lysine homopolymer hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
D-Lys-(D-Lys)n-D-Lys · xHBr
CAS Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

Poly-D-lysine hydrobromide, mol wt 1,000-5,000

form

powder or solid

mol wt

1,000-5,000

technique(s)

cell culture | mammalian: suitable

color

white to faint yellow

storage temp.

−20°C

SMILES string

O=C(C)[C@@](NC)([H])CCCCN.[Br]

InChI

1S/C6H14N2O2.BrH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H

InChI key

MEXAGTSTSPYCEP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly-D-lysine polymers can be used in preparing surfaces for cell attachment. The D-lysine polymers can also be used with cells that digest poly-L-lysine polymers and cause an excessive uptake of L-lysine.

This product is recommended as a cell culture substratum when using 0.5 - 1.0 mL of a 0.1 mg/mL solution to coat 25 cm2. Lower molecular weight versions of the product are less viscous, but high more molecular weight versions provide more attachment sites per molecule.

Biochem/physiol Actions

Poly-D-lysine (PDL) hydrobromide is a nonspecific attachment factor for cells useful in promoting cell adhesion to solid substrates by enhancing electrostatic interaction between negatively charged ions of the cell membrane and the culture surface. After absorption to the culture surface, poly-D-lysine increases the number of positively charged cell binding sites.

Components

Poly-D-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue. The hydrobromide allows the poly-D-lysine to be in a crystalline form soluble in water. A small amount of product may be found in the ß structure because the HBr interferes with hydrogen bonding between amino and either the carboxyl groups or N or O containing moieties.

Caution

Sterile solutions are stable for up to 2 years when stored at 2-8°C. It should be stored desiccated at -20°C.

Analysis Note

This product has a molecular weight of 1,000-4,000. To remove the HBr, dissolve this product in a neutral buffer and dialyze to remove the salts. In general, to use this product as an attachment factor, add 50 mL of sterile tissue culture grade water to 5 mg of poly-lysine, and aseptically coat the surface with 1 mL per 25 cm2 of solution. After 5 minutes, remove the solution through aspiration and thoroughly rinse the surface. Let dry for two hours before introducing cells and medium.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuan Zhang et al.
Autophagy, 12(9), 1538-1559 (2016-07-28)
BBC3 (BCL2 binding component 3) is a known apoptosis inducer; however, its role in microglial survival remains poorly understood. In addition to the classical transcription factor TRP53, Mir143 is involved in BBC3 expression at the post-transcriptional level. Here, we identify
Yunfei Chen et al.
Aging, 12(18), 18274-18296 (2020-09-24)
Traumatic brain injury (TBI) is a leading cause of morbidity and mortality in young individuals worldwide. There is currently no effective clinical treatment for TBI, but mesenchymal stem cell-derived exosomes have exhibited promising therapeutic effects. In this study, we performed
Qi Li et al.
Acta neuropathologica communications, 7(1), 217-217 (2019-12-25)
Perioperative sleep disturbance is a risk factor for persistent pain after surgery. Clinical studies have shown that patients with insufficient sleep before and after surgery experience more intense and long-lasting postoperative pain. We hypothesize that sleep deprivation alters L-type calcium
Yanhui Cai et al.
iScience, 23(5), 101136-101136 (2020-05-24)
Astrocytic glycogen is an important energy reserve in the brain and is believed to supply fuel during energy crisis. However, the pattern of glycogen metabolism in ischemic stroke and its potential therapeutic impact on neurological outcomes are still unknown. Here
Roberto de la Rica et al.
Analytical chemistry, 81(24), 10167-10171 (2009-11-17)
While cancer is still an implacable disease, many cancers can be cured if they are diagnosed in an early stage. Recently, it was reported that the transformation from normal cells to cancer cells can change their mechanoelastic properties to become

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service