Skip to Content
Merck
All Photos(1)

Documents

P4439

Sigma-Aldrich

Phosphatase, Alkaline from porcine kidney

lyophilized powder, ≥100 DEA units/mg protein

Synonym(s):

Orthophosphoric-monoester phosphohydrolase (alkaline optimum)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
42010105
NACRES:
NA.54

form

lyophilized powder

specific activity

≥100 DEA units/mg protein

composition

Protein, ≥50%

storage temp.

−20°C

Application

Alkaline phosphatase is used for conjugation to antibodies and other proteins for ELISA, Western blotting, and histochemical detection. Alkaline phosphatase is also used to prevent DNA self ligation and for radiolabeling.

Biochem/physiol Actions

Alkaline phosphatase is a hydrolase enzyme that dephosphorylates molecules such as nucleotides, proteins and alkaloids.

Packaging

Package size based on DEA units

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrophenyl phosphate per min at pH 9.8 at 37 °C.

Physical form

Lyophilized fromTris-HCl buffer, with magnesium chloride and zinc chloride, pH 8.0

Analysis Note

Protein determined by biuret.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sequencing end-labeled DNA with base-specific chemical cleavages.
A M Maxam et al.
Methods in enzymology, 65(1), 499-560 (1980-01-01)
Naomi Ofer et al.
The Journal of biological chemistry, 288(16), 11287-11293 (2013-03-09)
Protein degradation via prokaryotic ubiquitin-like protein (Pup) tagging is conserved in bacteria belonging to the phyla Actinobacteria and Nitrospira. The physiological role of this novel proteolytic pathway is not yet clear, although in Mycobacterium tuberculosis, the world's most threatening bacterial
Gouri Chaudhuri et al.
Indian journal of biochemistry & biophysics, 50(1), 64-71 (2013-04-27)
The hydrolysis of p-nitrophenyl phosphate (pNPP) by calf intestinal alkaline phosphatase (CIAP) was investigated with respect to kinetic parameters such as V(max), K(m) and K(cat) under varying pH, buffers, substrate concentration, temperature and period of incubation. Highest activity was obtained
Lars Hansen et al.
American journal of human genetics, 92(4), 575-583 (2013-04-09)
PGAP2 encodes a protein involved in remodeling the glycosylphosphatidylinositol (GPI) anchor in the Golgi apparatus. After synthesis in the endoplasmic reticulum (ER), GPI anchors are transferred to the proteins and are remodeled while transported through the Golgi to the cell
Insulin affects dopamine metabolism in Drosophila females under normal and stress conditions.
N V Adonyeva et al.
Doklady. Biochemistry and biophysics, 448, 40-42 (2013-03-13)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service