Skip to Content
Merck
All Photos(2)

Key Documents

P3752

Sigma-Aldrich

Phosphatase, Acid from potato

lyophilized powder, ≥0.5 unit/mg solid

Synonym(s):

(Orthophosphoric-monoester phosphohydrolase acid optimum), Acid Phosphatase from potato

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
42010102
NACRES:
NA.54

biological source

potato

Quality Level

form

lyophilized powder

specific activity

≥0.5 unit/mg solid

mol wt

69 kDa

application(s)

diagnostic assay manufacturing

storage temp.

−20°C

SMILES string

O(CC(O)C)CC#C

InChI

1S/C6H10O2/c1-3-4-8-5-6(2)7/h1,6-7H,4-5H2,2H3

InChI key

GZCWLCBFPRFLKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acid phosphatase is routinely used to dephosphorylate proteins and nucleic acids. Acid phosphatase is an acid hydrolase that normally resides in lysosomes. It is used as marker for the identification of lysosomes in subcellular fractionations. Acid phosphatase, product P3752, has been used as a control in order to study how Mycobacterium bovis BCG effects the levels of acid phosphatase activity in alveolar macrophages.

Biochem/physiol Actions

Acid phosphatase is a cytoplasmic lysosomal hydrolase that catalyzes the conversion of an orthophosphoric monoester and water to an alcohol and orthophosphate.

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrophenyl phosphate per min at pH 4.8 at 37 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Zhang et al.
Biosensors & bioelectronics, 141, 111442-111442 (2019-06-30)
Phenol contains an exchangeable hydroxyl proton resonant at 4.8 ppm from the resonance frequency of water in the 1H nuclear magnetic resonance (1H NMR) spectrum, enabling itself to be detected at sub-mM concentration by either chemical exchange saturation transfer magnetic resonance
Julia R Smith et al.
Physiological genomics, 31(2), 357-363 (2007-08-09)
We have developed a method to determine the degree of phosphorylation of a peptide in a complex mixture without enrichment or operation of the mass spectrometer in negative ion mode. Yeast lysate containing known amounts of synthetic peptides (VPQLEIVPNSAEERLHSMK and
K Zyła et al.
The British journal of nutrition, 74(1), 3-17 (1995-07-01)
An in vitro method was developed to predict inorganic P release from maize-soyabean poultry feeds containing supplemental phytase (EC 3.1.3.8), and to quantify the effect of acid phosphatase (EC 3.1.3.2), fungal protease (EC 3.4.23.6) and Aspergillus niger cellulase (EC 3.2.1.4)
I N Mbawuike et al.
Infection and immunity, 51(2), 483-489 (1986-02-01)
The kinetics of induction of the bronchoalveolar cell population (i.e., alveolar macrophages [AM], lymphocytes, and polymorphonuclear leukocytes) was studied in mice inoculated intravenously with heat-killed Mycobacterium bovis BCG. Injection of BCG at 100 and 500 micrograms but not at 10
Filipe Natalio et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(21), 4973-4980 (2017-03-02)
Boron's unusual properties inspired major advances in chemistry. In nature, the existence and importance of boron has been fairly explored (e.g. bacterial signaling, plant development) but its role as biological catalyst was never reported. Here, we show that boric acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service