P4126
Phe-Phe
≥98% (TLC)
Synonym(s):
L-Phenylalanyl-L-phenylalanine, Di-L-phenylalanine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5CH2CH(NH2)CONHCH(CH2C6H5)COOH · H2O
CAS Number:
Molecular Weight:
312.36 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Product Name
Phe-Phe,
Assay
≥98% (TLC)
form
powder
color
white
storage temp.
−20°C
SMILES string
N[C@@H](Cc1ccccc1)C(=O)N[C@@H](Cc2ccccc2)C(O)=O
InChI
1S/C18H20N2O3/c19-15(11-13-7-3-1-4-8-13)17(21)20-16(18(22)23)12-14-9-5-2-6-10-14/h1-10,15-16H,11-12,19H2,(H,20,21)(H,22,23)/t15-,16-/m0/s1
InChI key
GKZIWHRNKRBEOH-HOTGVXAUSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Supramolecular nanofibers and hydrogels of nucleopeptides.
Xinming Li et al.
Angewandte Chemie (International ed. in English), 50(40), 9365-9369 (2011-09-29)
Robert C Dunbar et al.
Journal of the American Chemical Society, 133(5), 1212-1215 (2011-01-05)
Chirality reversal of a residue in a peptide can change its mode of binding to a metal ion, as shown here experimentally by gas-phase IR spectroscopy of peptide-metal ion complexes. The binding conformations of Li(+), Na(+), and H(+) with the
H Mozsolits et al.
Biophysical journal, 77(3), 1428-1444 (1999-08-31)
The interaction of three bioactive peptides, bombesin, beta-endorphin, and glucagon with a phosphatidylcholine monolayer that was immobilized to porous silica particles and packed into a stainless steel column cartridge, has been studied using dynamic elution techniques. This immobilized lipid monolayer
Gabriel Riquelme et al.
Metabolites, 10(10) (2020-10-22)
Preprocessing data in a reproducible and robust way is one of the current challenges in untargeted metabolomics workflows. Data curation in liquid chromatography-mass spectrometry (LC-MS) involves the removal of biologically non-relevant features (retention time, m/z pairs) to retain only high-quality
Meital Reches et al.
Physical biology, 3(1), S10-S19 (2006-04-04)
Molecular self-assembly offers new routes for the fabrication of novel materials at the nano-scale. Peptide-based nanostructures represent nano-objects of particular interest, as they are biocompatible, can be easily synthesized in large amounts, can be decorated with functional elements and can
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service