Skip to Content
Merck
All Photos(1)

Key Documents

327204

Sigma-Aldrich

Paraffin wax

mp 53-58 °C (ASTM D 87)

Synonym(s):

Paraffin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

form

beads
waxy solid

mp

53-58 °C (ASTM D 87)

InChI key

JWHAUXFOSRPERK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Paraffin Wax is a solidified microcrystal that is produced by continuous boiling of residual oils that include iso-alkanes. It is characterized in terms of soft and hard paraffins with a melting point in the range of 45-60°C.[1]

Application

Paraffin wax can be used as a phase changing material (PCM) that can be loaded with exfoliated graphite nanoplatelets (xGnP) for the development of energy saving building materials.[2] It may be used as a combustion material for hybrid rocket motor applications.[3] Paraffin wax/graphite nanoparticles based nanocomposite may be used in the fabrication of hybrid storage material for solar still to desalinate seawater.[4]

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

602.6 °F - Pensky-Martens closed cup

Flash Point(C)

317 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

<SMALL>Bancroft JD</SMALL>
Theory and Practice of Histotechnology null
Effect of asphaltenes on equilibrium and rheological properties of waxy model systems
Luis Alberto Alcazar-Vara, et al.
Fuel: The Science and Technology of Fuel and Energy, 93, 200-212 (2012)
Paraffin products: properties, technologies, applications, 14 (1982)
Improvement of oil flowability by assembly of comb-type copolymers with paraffin and asphaltene
Li L, et al.
AIChE Journal, 58(7), 2254-2261 (2012)
Effect of graphite mass concentrations in a mixture of graphite nanoparticles and paraffin wax as hybrid storage materials on performances of solar still
Kabeel AE, et al.
Renewable Energy, 132, 119-128 (2019)

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What is the latent heat capacity of this product?

    1 answer
    1. The thermophysical properties are not evaluated for this product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service