Skip to Content
Merck

Skip To

A5597

Xylene Substitute

greener alternative

Synonym(s):

for microscopy

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12171500
EC Number:
265-068-8
NACRES:
NA.47
Form:
solution
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

density

0.723 g/cm3

application(s)

hematology
histology

greener alternative category

storage temp.

2-8°C

General description

Compatible with all tissue processors. May not be compatible with all permanent mounting media.

Application

Xylene Substitute has been used to dewax or de-paraffinize the specimen sections after the staining process.We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to the standard use of formalin for tissue processing. For more information see www.sigmaaldrich.com/catalog/papers/19608083.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
784751.09843PHR1313
density

0.723 g/cm3

density

0.75 g/mL at 20 °C

density

0.78-0.795 g/cm3 at 20 °C

density

0.868 g/mL at 25 °C (lit.)

form

solution

form

liquid

form

liquid

form

-

storage temp.

2-8°C

storage temp.

-

storage temp.

15-25°C

storage temp.

2-30°C

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

greener alternative product characteristics

-

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

greener alternative product characteristics

-

sustainability

Greener Alternative Product

sustainability

-

sustainability

Greener Alternative Product

sustainability

-

application(s)

hematology
histology

application(s)

-

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

pharmaceutical (small molecule)


Still not finding the right product?

Explore all of our products under Xylene Substitute


pictograms

FlameHealth hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

flash_point_f

63.0 °F - closed cup

flash_point_c

17.2 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters