Skip to Content
Merck
All Photos(4)

Key Documents

P0380

Sigma-Aldrich

L-Proline

≥99% (HPLC), ReagentPlus®

Synonym(s):

(S)-Pyrrolidine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
Beilstein:
80810
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Proline, ReagentPlus®, ≥99% (HPLC)

Quality Level

product line

ReagentPlus®

Assay

≥99% (HPLC)

form

powder

impurities

L-Hydroxyproline, free

color

white

mp

228 °C (dec.) (lit.)

application(s)

detection

SMILES string

OC(=O)[C@@H]1CCCN1

InChI

1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)/t4-/m0/s1

InChI key

ONIBWKKTOPOVIA-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Proline has been used:
  • As a supplement during the preparation of chondrogenic medium and synthetic dextrose minimal medium (SD).
  • As a standard during the identification of metabolites in serum samples.
  • In a study to prepare L-proline-L-phenylalanine (L-Pro-L-Phe) mixture in aqueous acetonitrile.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Andrew Davison
Eureka: Biochemistry & Metabolism, 2015, 76-76 (2015)
Mieczysław Sajewicz et al.
Journal of chromatographic science, 53(1), 31-37 (2014-03-05)
We employ the achiral liquid chromatography with diode array, evaporative light scattering and mass spectrometric detection (HPLC-DAD, HPLC-ELSD and LC-MS) to assess structural instability (understood as spontaneous oscillatory chiral conversion and spontaneous oscillatory condensation) of the two pairs of amino
Proline: a multifunctional amino acid
La' szlo' Szabados and Arnould Savoure
Trends in Plant Science, 1-9 (2009)
Metabolomic insights into system-wide coordination of vertebrate metamorphosis.
Ichu TA, et al.
BMC Developmental Biology, 14(1), 1-1 (2014)
Natasja Leth Joergensen et al.
BioResearch open access, 4(1), 109-114 (2015-08-27)
Cartilage regeneration is a fast growing field that combines biotechnology and molecular techniques in creating new tissue mimicking the native microenvironment. Human embryonic stem cells (hESCs) are a highly potent cell source for cartilage regeneration owing to their infinite proliferation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service