Skip to Content
Merck
All Photos(2)

Key Documents

P2850

Sigma-Aldrich

Phosphatase Inhibitor Cocktail 1

DMSO solution, mixture of inhibitors

Synonym(s):

Phosphatase Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200
NACRES:
NA.54

Product Name

Phosphatase Inhibitor Cocktail 1, DMSO solution

form

DMSO solution

Quality Level

shipped in

wet ice

storage temp.

2-8°C

General description

Phosphatase Inhibitor Cocktail 1 includes a mixture of inhibitors that will inhibit the L-isozymes of alkaline phosphatase as well as serine/threonine protein phosphatases such as PP1 and PP2A. This product contains microcystinLR, cantharidin, and (−)-p-bromotetramisole. Phosphatase Inhibitor Cocktail 1 has been replaced with P0044.

Application

Phosphatase Inhibitor Cocktail 1 has been used in nuclear protein isolation and electrophoretic mobility shift assay (EMSA) for catenin activation. It has also been used in western blot analysis.

Biochem/physiol Actions

Phosphatases are active regulators of a number of important biological processes. Inhibition of phosphatases by small molecules is useful to study its biological role.

Quantity

Recommended usage is a 100-fold dilution.

Analysis Note

Tested on cell extracts from animal tissues such as bovine liver, human placenta, and rabbit muscle, as well as on extracts from A431 and Jurkat cell lines.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Class III β-tubulin expression and in vitro resistance to microtubule targeting agents.
Stengel C, et al.
British Journal of Cancer, 102(2), 316-316 (2010)
Larger therapeutic window for steroid versus VEGF-A inhibitor in inflammatory angiogenesis: surprisingly similar impact on leukocyte infiltration.
Nakao S, et al.
Investigative Ophthalmology & Visual Science, 53(7), 3296-3302 (2012)
X Zhao et al.
Mucosal immunology, 10(6), 1553-1568 (2017-04-13)
Virulent Mycobacterium tuberculosis (Mtb) triggers necrosis in host Mϕ, which is essential for successful pathogenesis in tuberculosis. Here we demonstrate that necrosis of Mtb-infected Mϕ is dependent on the action of the cytosolic Receptor Interacting Protein Kinase 3 (RIPK3) and
Discovery of protein phosphatase inhibitor classes by biology-oriented synthesis.
Noren M A, et al.
Proceedings of the National Academy of Sciences of the USA, 103(28), 10606-10611 (2006)
Aberrant Wnt/β-catenin signaling in pancreatic adenocarcinoma.
Zeng G, et al.
Neoplasia, 8(4), 279-289 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service