Skip to Content
Merck
All Photos(1)

Key Documents

C-008

Supelco

Cocaine solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO4
CAS Number:
Molecular Weight:
303.35
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IB (Portugal)

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

COC(=O)[C@H]1[C@H](C[C@@H]2CC[C@H]1N2C)OC(=O)c3ccccc3

InChI

1S/C17H21NO4/c1-18-12-8-9-13(18)15(17(20)21-2)14(10-12)22-16(19)11-6-4-3-5-7-11/h3-7,12-15H,8-10H2,1-2H3/t12-,13+,14-,15+/m0/s1

InChI key

ZPUCINDJVBIVPJ-LJISPDSOSA-N

General description

A Certified Spiking Solution® suitable for use in LC/MS or GC/MS testing applications for forensic analysis, urine drug testing, or clinical toxicology. Cocaine is a tropane alkaloid and illicit drug obtained from the leaves of the coca plant. This popular recreational drug is a powerful stimulant of the central nervous system with appetite suppressant and topical anesthetic properties.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coca science seeks an answer in kilos.
Antonio Regalado
Science (New York, N.Y.), 341(6145), 453-453 (2013-08-03)
Carlos Manuel Lima Reis da Silva et al.
Journal of psychoactive drugs, 45(2), 195-198 (2013-08-03)
The topic of ophthalmic use of cocaine in clinical practice has a long history; nevertheless, the possible influence and pathways of action of inhaled cocaine in the human eye remain unknown. This study evaluates the effect of snorted cocaine in
Watchareewan Thongsaard et al.
Journal of the Medical Association of Thailand = Chotmaihet thangphaet, 96 Suppl 1, S85-S89 (2013-06-04)
Thunbergia laurifolia Linn. (TL) is a herbal medicine used as an antidote for several poisonous agents in Thai traditional medicine. TL were reported not only to significantly increase potassium-stimulated dopamine release from rat striatal slices but also potentiated the effect
Thomas C Jhou et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(17), 7501-7512 (2013-04-26)
Many strong rewards, including abused drugs, also produce aversive effects that are poorly understood. For example, cocaine can produce aversive conditioning after its rewarding effects have dissipated, consistent with opponent process theory, but the neural mechanisms involved are not well
Wei Xu et al.
Life sciences, 92(23), 1101-1109 (2013-04-30)
Chronic administration of cocaine attenuates delta opioid receptor (DOPR) signaling in the striatum and the desensitization is mediated by the indirect actions of cocaine on dopamine D1 receptors (D1R). In addition, DOPR and D1R co-exist in some rat striatal neurons.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service