Skip to Content
Merck
All Photos(1)

Key Documents

P1784

Sigma-Aldrich

Pentoxifylline

solid

Synonym(s):

3,7-Dimethyl-1-(5-oxohexyl)xanthine, Trental

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18N4O3
CAS Number:
Molecular Weight:
278.31
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

color

white

solubility

H2O: ≥43 mg/mL, colorless to almost colorless

originator

Sanofi Aventis

SMILES string

CN1C=NC2C1C(=O)N(CCCCC(C)=O)C(=O)N2C

InChI

1S/C13H20N4O3/c1-9(18)6-4-5-7-17-12(19)10-11(14-8-15(10)2)16(3)13(17)20/h8,10-11H,4-7H2,1-3H3

InChI key

MQGNNJQTNFHNHK-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Pentoxifylline has been used:
  • used in the combinatorial treatment with itraconazole for paracoccidioidomycosis (PCM)
  • to treat harvested sperms to check the effect of ′pentoxifylline exposed sperms′ in the contribution of embryonic growth
  • to intrathecally inject female and male mice to investigate whether astrocytes and microglia could be causally involved in the maintenance of pain-like behaviour

Biochem/physiol Actions

Pentoxifylline (PTX) is considered as a nonspecific phosphodiesterase inhibitor. It possesses rheologic properties. Pentoxifylline is used to treat peripheral vascular disease. It has the ability to block the phosphorylation of I kappa B-alpha (IĸBα) in serines 32 and 36.
Phosphodiesterase inhibitor; inhibits synthesis of tumor necrosis factor α (TNF-α).

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Histamine Synthesis and Metabolism and Phosphodiesterases pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Lact.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PP-023-Effect of ICSI with `pentoxifylline exposed sperms? on subsequent embryo development
Mangoli V S, et al.
Reproductive Biomedicine Online, 32, S11-S11 (2016)
B K Reuter et al.
The American journal of physiology, 277(4 Pt 1), G847-G854 (1999-10-12)
Although the ability of nonsteriodal anti-inflammatory drugs (NSAIDs) to injure the small intestine has been well established in humans and animals, the mechanism involved in this type of injury has yet to be elucidated. The cytokine tumor necrosis factor-alpha (TNF-alpha)
Maria Del Mar Castro et al.
Pathogens (Basel, Switzerland), 11(3) (2022-03-27)
Addition of the immunomodulator pentoxifylline (PTX) to antimonial treatment of mucosal leishmaniasis has shown increased efficacy. This randomized, double-blind, placebo-controlled trial evaluated whether addition of pentoxifylline to meglumine antimoniate (MA) treatment improves therapeutic response in cutaneous leishmaniasis (CL) patients. Seventy-three
Combined itraconazole-pentoxifylline treatment promptly reduces lung fibrosis induced by chronic pulmonary paracoccidioidomycosis in mice
Naranjo T W, et al.
Pulmonary Pharmacology & Therapeutics, 24(1), 81-91 (2011)
Teresa Fernandez-Zafra et al.
Pain, 160(1), 224-236 (2018-09-25)
Recent studies have suggested a sexually dimorphic role of spinal glial cells in the maintenance of mechanical hypersensitivity in rodent models of chronic pain. We have used the collagen antibody-induced arthritis (CAIA) mouse model to examine differences between males and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service