Skip to Content
Merck
All Photos(5)

Key Documents

T1633

Sigma-Aldrich

Theophylline

≥99% (HPLC), powder, phosphodiesterase inhibitor

Synonym(s):

1,3-Dimethylxanthine, 2,6-Dihydroxy-1,3-dimethylpurine, 3,7-Dihydro-1,3-dimethyl-1H-purine-2,6-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N4O2
CAS Number:
Molecular Weight:
180.16
Beilstein:
13463
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Product Name

Theophylline, anhydrous, ≥99%, powder

grade

anhydrous

Assay

≥99%

form

powder

color

white

solubility

H2O: slightly soluble 8.3 mg/ml
NH4OH: 50 mg/ml, clear, colorless
alcohol: 12.5 mg/ml
chloroform: soluble 9.1 mg/ml
0.1 M HCl: soluble
0.1 M NaOH: soluble
ammonium hydroxide: soluble
aqueous base: soluble
diethyl ether: slightly soluble
dilute HCl: soluble
dilute nitric acid: soluble
ethanol: moderately soluble

originator

Forest Labs

SMILES string

CN1C(=O)N(C)c2[nH]cnc2C1=O

InChI

1S/C7H8N4O2/c1-10-5-4(8-3-9-5)6(12)11(2)7(10)13/h3H,1-2H3,(H,8,9)

InChI key

ZFXYFBGIUFBOJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Theophylline has been used to study its effects on rat gene expression in the ubiquitin-proteasome pathway that regulates spermatogenesis and epididymal sperm quality. Theophylline has also been used as an internal standard for the measurement of plasma paracetamol levels in humans.

Biochem/physiol Actions

Phosphodiesterase inhibitor; diuretic; cardiac stimulant; muscle relaxant; asthma medication.
Theophylline is a tea alkaloid that functions as a nonselective PDE4 inhibitor. It can induce smooth muscle relaxation in the bronchiolae of asthma patients. At increased concentrations, theophylline can function as a reprotoxic agent and can cause infertility by incapacitating Sertoli cells. This subsequently causes the premature release of late differentiating spermatogenic cells.

Features and Benefits

This compound is a featured product for ADME Tox and Cyclic Nucleotide research. Discover more featured ADME Tox and Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Adenosine Receptors and Phosphodiesterases pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Forest Labs. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

Theophylline dissolves in 1 M NH4OH at 50 mg/ml to yield a clear, colorless solution. It is soluble in 0.1 M HCl, 0.1 M NaOH and is slightly soluble(8.3 mg/ml) in water. It is also moderately soluble in ethanol. Furthermore, it is soluble in alcohol (12.5 mg/ml), and chloroform (9.1 mg/ml), alkali hydroxides, ammonia, dilute hydrochloric or nitric acid, but is sparingly soluble in ether.

The solubility of the methylxanthines is low, but can be enhanced by the formation of complexes (usually 1:1) with a wide variety of compounds such as ethylenediamine (to form aminophylline). The formation of complex double salts (caffeine and sodium benzoate) or true salts (like choline theophyllinate, and oxtriphylline) also improves aqueous solubility. These salts or complexes dissociate to yield the parent methylxanthines when dissolved in biological fluids and should not be confused with covalently modified derivatives such asdyphylline (1,3-dimethyl-7-(2,3-dihydroxypropyl)-xanthine).

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F J Sevilla-Tirado et al.
Methods and findings in experimental and clinical pharmacology, 25(7), 531-535 (2003-10-23)
The aim of this study was to compare the main pharmacokinetic characteristics of two new paracetamol formulations, powder sachet and tablet, with that of three commercially available paracetamol formulations: two conventional solid tablets and one effervescent tablet. Twelve healthy volunteers
Nagi Reddy Dumpa et al.
Pharmaceutics, 12(1) (2020-01-16)
This study was performed to develop novel core-shell gastroretentive floating pulsatile drug delivery systems using a hot-melt extrusion-paired fused deposition modeling (FDM) 3D printing and direct compression method. Hydroxypropyl cellulose (HPC) and ethyl cellulose (EC)-based filaments were fabricated using hot-melt
Mohammed Al-Sharabi et al.
International journal of pharmaceutics, 584, 119380-119380 (2020-05-15)
Fast disintegrating tablets have commonly been used for fast oral drug delivery to patients with swallowing difficulties. The different characteristics of the pore structure of such formulations influence the liquid transport through the tablet and hence affect the disintegration time
Matthew T Eddy et al.
Structure (London, England : 1993), 29(2), 170-176 (2020-11-26)
In drug design, G protein-coupled receptor (GPCR) partial agonists enable one to fine-tune receptor output between basal and maximal signaling levels. Here, we add to the structural basis for rationalizing and monitoring partial agonism. NMR spectroscopy of partial agonist complexes
Wenjie Zhao et al.
Bio-protocol, 11(4), e3919-e3919 (2021-03-19)
In the last several years, as evidence of a surged number of GPCR-G complex structures, the expressions of GPCRs and G proteins for structural biology have achieved tremendous successes, mostly in insect and mammalian cell systems, resulting in more than

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service