Skip to Content
Merck
All Photos(1)

Key Documents

P0440

Sigma-Aldrich

Pimaricin preparation

~2.5% (γ-irradiated Pimaricin), aqueous suspension

Synonym(s):

Pimaricin, Tennecetin, Natamycin preparation

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H47NO13
CAS Number:
Molecular Weight:
665.73
Beilstein:
1614878
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

form

aqueous suspension

Quality Level

concentration

~2.5% (γ-irradiated Pimaricin)

solubility

DMSO: soluble

density

1.0 g/mL at 20 °C (lit.)

antibiotic activity spectrum

fungi
yeast

Mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

[H][C@]12C[C@@H](O[C@@H]3O[C@H](C)[C@@H](O)[C@H](N)[C@@H]3O)\C=C\C=C\C=C\C=C\C[C@@H](C)OC(=O)\C=C\[C@@]4([H])O[C@]4([H])C[C@H](O)C[C@](O)(C[C@H](O)[C@H]1C(O)=O)O2

InChI

1S/C33H47NO13/c1-18-10-8-6-4-3-5-7-9-11-21(45-32-30(39)28(34)29(38)19(2)44-32)15-25-27(31(40)41)22(36)17-33(42,47-25)16-20(35)14-24-23(46-24)12-13-26(37)43-18/h3-9,11-13,18-25,27-30,32,35-36,38-39,42H,10,14-17,34H2,1-2H3,(H,40,41)/b4-3+,7-5+,8-6+,11-9+,13-12+/t18-,19-,20+,21+,22+,23-,24-,25+,27-,28+,29-,30+,32+,33-/m1/s1

InChI key

NCXMLFZGDNKEPB-FFPOYIOWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is a saline suspension with approximately 2.5% pimaricin γ-irradiated.

Pimaricin is a polyene antifungal antibiotic produced by Streptomyces natalensis from soil near Pietermaritzburg, South Africa.1 Pimaricin has antimicrobial activity similar to that of nystatin. In addition, it is active against Trichomonas vaginalis. Pimaricin is used in the treatment of candidiasis, trichomoniasis, fungal keratitis and aspergillosis. It has also been used as a food additive in some countries. In some studies, it has been shown to decrease the amount of mold upon which the Dermatophagoides pteronyssinus (house-dust mite) is dependent.2

Application

Pimaricin is an amphoteric antibiotic from Streptomyces natalensis or S. chattanoogensis. It is used for a variety of fungal infections, mainly topically. It is used as an ergosterol and cholesterol binding agent to study lipid bilayer dynamics, especially in fungal cells. It is used to study pimaricin biosynthesis and as a fungicide in agar media.

Biochem/physiol Actions

An antifungal polyene macrolide that binds specifically to ergosterol and blocks fungal growth. However, unlike nystatin and filipin, pimaricin does not change the permeability of the plasma membrane.

Preparation Note

The product can be further diluted with 0.5 M sodium chloride giving a suspension as well. The product can also be further diluted into an organic solvent such as dimethylformamide (may also be soluble in DMSO).

The product and any aqueous dilutions will be suspensions and should not be sterile filtered.

Storage and Stability

The product is not degraded by a single freeze-thaw cycle. The diluted product should be aliquoted and stored frozen.

Other Notes

20ml
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.Light Sensitive

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eliseo Recio et al.
The Journal of biological chemistry, 279(40), 41586-41593 (2004-07-03)
A chemically novel autoinducer (PI factor) has been purified from cultures of the pimaricin producer Streptomyces natalensis ATCC27448. The chemical structure of the PI molecule was identified as 2,3-diamino-2,3-bis (hydroxymethyl)-1,4-butanediol. Pimaricin biosynthesis in S. natalensis npi287, a mutant impaired in
J C Pedersen
Applied and environmental microbiology, 58(3), 1064-1066 (1992-03-01)
Fungal inhibition in four commonly used agar media was improved by substituting natamycin (pimaricin) for cycloheximide. The recovery of bacteria was not affected by natamycin, whereas fungal contamination from a variety of samples was significantly suppressed. Furthermore, natamycin lacks the
Namrata Sharma et al.
Ophthalmology, 120(4), 677-681 (2012-12-19)
To compare the efficacy of topical voriconazole and topical natamycin with that of intrastromal voriconazole and topical natamycin in patients with recalcitrant fungal keratitis. Randomized clinical trial. Forty eyes of 40 patients with fungal keratitis (positive smear or culture results
Pranab K Mukherjee et al.
Investigative ophthalmology & visual science, 53(8), 4450-4457 (2012-06-07)
Fusarium is a major cause of microbial keratitis, and its ability to form biofilms was suggested as a contributing factor in recent outbreaks. We investigated the ability of outbreak Fusarium isolates (F. solani species complex [FSSC] and F. oxysporum species
R S Bhatta et al.
International journal of pharmaceutics, 432(1-2), 105-112 (2012-05-10)
The aim of this study was to prepare natamycin encapsulated lecithin/chitosan mucoadhesive nanoparticles (NPs) for prolonged ocular application. These NPs were characterized by their mean particle size 213nm, encapsulation efficiency 73.57%, with a theoretical drug loading 5.09% and zeta potential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service