Skip to Content
Merck

Skip To

P1024

Poly-D-lysine hydrobromide

suitable for cell culture, Mol wt ≥300,000

Synonym(s):

D-Lysine homopolymer hydrobromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
D-Lys-(D-Lys)n-D-Lys · xHBr
CAS Number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Poly-D-lysine hydrobromide, mol wt ≥300,000

form

powder or solid

Quality Segment

mol wt

≥300,000

technique(s)

cell culture | mammalian: suitable

color

white to off-white

storage temp.

−20°C

SMILES string

O=C(C)[C@@](NC)([H])CCCCN.[Br]

InChI

1S/C6H14N2O2.BrH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H

InChI key

MEXAGTSTSPYCEP-UHFFFAOYSA-N

Application

Poly-D-lysine polymers can be used in preparing surfaces for cell attachment. The D-lysine polymers can also be used with cells that digest poly-L-lysine polymers and cause an excessive uptake of L-lysine.

This product is recommended as a cell culture substratum when using 0.5 - 1.0 mL of a 0.1 mg/mL solution to coat 25 cm2. Lower molecular weight versions of the product are less viscous, but high more molecular weight versions provide more attachment sites per molecule.

Biochem/physiol Actions

Poly-D-lysine (PDL) hydrobromide is a nonspecific attachment factor for cells useful in promoting cell adhesion to solid substrates by enhancing electrostatic interaction between negatively charged ions of the cell membrane and the culture surface. After absorption to the culture surface, poly-D-lysine increases the number of positively charged cell binding sites.

Preparation Note

This product has a molecular weight >300,000. To remove the HBr, dissolve this product in a neutral buffer and dialyze to remove the salts. In general, to use this product as an attachment factor, add 50 mL of sterile tissue culture grade water to 5 mg of poly-lysine, and aseptically coat the surface with 1 mL per 25 cm2 of solution. After 5 minutes, remove the solution through aspiration and thoroughly rinse the surface. Let dry for two hours before introducing cells and medium.

Other Notes

Poly-D-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue. The hydrobromide allows the poly-D-lysine to be in a crystalline form soluble in water. A small amount of product may be found in the β structure because the HBr interferes with hydrogen bonding between amino and either the carboxyl groups or N or O containing moieties.

Disclaimer

Sterile solutions are stable for up to 2 years when stored at 2-8°C. It should be stored desiccated at -20°C.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P7886P0899P7405
form

powder or solid

form

solid

form

solid

form

lyophilized powder

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Quality Level

300

Quality Level

200

Quality Level

200

Quality Level

200

color

white to off-white

color

white to off-white

color

white to off-white

color

-

mol wt

≥300,000

mol wt

30,000-70,000

mol wt

70,000-150,000

mol wt

>300,000


Still not finding the right product?

Explore all of our products under Poly-D-lysine hydrobromide


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. What type of Poly-D-lysine hydrobromide is most suitable for cell attachment?

    1 answer
    1. Poly-D-lysine hydrobromide with a molecular weight greater than 30,000 Da is recommended for cell attachment. The poly-D-lysine hydrobromide with a molecular weight of 84,000 Da has more attachment sites per polymer chain available to the cells. This type is most suitable for cell attachment.

      Helpful?

Reviews

No rating value

Active Filters