Skip to Content
Merck
All Photos(1)

Key Documents

79390

Sigma-Aldrich

Phosphatase, Alkaline from calf intestinal mucosa

suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)

Synonym(s):

Alkaline Phosphatase from calf intestinal mucosa

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
42010105
NACRES:
NA.54

biological source

bovine (calf) intestinal mucosa

form

solution (clear, colorless)

specific activity

~2500 U/mg protein (~10 mg/ml)

mol wt

Mr ~140000

technique(s)

enzyme immunoassay: suitable

impurities

0.1 mM ZnCl2
1 mM MgCl2
3 M NaCl
30 mM triethanolamine

pH

~7.6

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Alkaline phosphatase is used in the laboratory to remove phosphate monoesters which prevents self-ligation. It is also used in the dairy industry to confirm pasturization of milk. Alkaline phosphatse is commonly conjugated to antibodies and used in immunoblotting, ELISA′s, and other antibody based detection assays.

Biochem/physiol Actions

Alkaline phosphatase is a hydrolytic enzyme which dephosphorylates a variety of molecules under basic conditions.

Unit Definition

1 U corresponds to the amount of enzyme which hydrolyzes 1 μmol 4-nitrophenyl phosphate per minute at pH 9.8 and 37 °C

Reconstitution

The solution can be used without dialysis (for enzyme immuno-assay).

Other Notes

Sales restrictions may apply
Used in highly sensitive (pg/m) enzyme immunoassays with LCEC detection of phenol; Comparison of three methods for conjugating alkaline phosphatase to human placental lactogen; Preparation of enzyme-antibody conjugates; Liposome immunoassay with antigen-coupled liposomes containing alkaline phosphatase

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C K Kim et al.
Journal of immunological methods, 159(1-2), 101-106 (1993-02-26)
Immunoliposomes were prepared and their immunoassay applications investigated. Liposomes were prepared from cholesterol and phospholipids including maleimidobenzoylphosphatidylethanolamine (MBPE) for conjunction with thiol-containing antigens. Alkaline phosphatase (APase) was entrapped in the liposome and BSA, the antigen, was modified by reaction with
Opioid receptor agonists enhance the phosphorylation state of Fas-associated death domain (FADD) protein in the rat brain: functional interactions with casein kinase Ialpha, Galpha(i) proteins, and ERK1/2 signaling.
Garcia-Fuster, M.J., et al.
Neuropsychopharmacology, 55, 886-899 (2008)
M.I. Halliday et al.
Biochemical Society Transactions, 14, 473-473 (1986)
K.R. Wehrmeyer et al.
Journal of Liquid Chromatography, 6, 2141-2141 (1983)
D G Williams
Journal of immunological methods, 72(1), 261-268 (1984-08-03)
One of the requirements for enzyme immunoassay is the formation of a labelled component of the assay system which can be either the ligand or the binder. Three methods for conjugating alkaline phosphatase to human placental lactogen were investigated, involving

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service