P1941
Poly(D,L-lactide-co-glycolide)
lactide:glycolide (75:25), mol wt 66,000-107,000
Synonym(s):
LACTEL® B6007-1, PLGA
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
[C3H4O2]x[C2H2O2]y
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
form
pellets
Quality Level
feed ratio
lactide:glycolide (75:25)
mol wt
66,000-107,000
storage temp.
−20°C
SMILES string
OCC(O)=O.CC(O)C(O)=O
InChI key
XBBVURRQGJPTHH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
The product is an amorphous solid with a transition temperature (Tg) in the range of 45-50°C. It is soluble in most organic solvents such as methylene chloride, tetrahydrofuran, ethyl acetate, acetone, chloroform, and hexafluoroispropanol.
Application
Poly(D,L-lactide-co-glycolide) is a synthetic amino acid that has been used to deliver macromolecular therapeutics.
Features and Benefits
Controlled release of bioactive agents, sutures and bioabsorbable implantable devices.
Physical form
Biocompatible, biodegradable polymer.
Legal Information
Lactel is a registered trademark of Evonik Corp
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Fabrication of amorphous strontium polyphosphate microparticles that induce mineralization of bone cells in vitro and in vivo
Werner E.G.Muller
Acta Biomaterialia, 50, 89-101 (2017)
Robert M Stefani et al.
Acta biomaterialia, 102, 326-340 (2019-12-06)
Articular cartilage defects are a common source of joint pain and dysfunction. We hypothesized that sustained low-dose dexamethasone (DEX) delivery via an acellular osteochondral implant would have a dual pro-anabolic and anti-catabolic effect, both supporting the functional integrity of adjacent
Rongcai Liang et al.
International journal of pharmaceutics, 454(1), 344-353 (2013-07-23)
Peptide or protein degradation often occurs when water flows into the dosage form. The aim of this study was to investigate the effect of water on exenatide acylation in poly(lactide-co-glycolide) (PLGA) microspheres. Exenatide-loaded PLGA microspheres were incubated at different relative
Shu-Chun Chuang et al.
Parasites & vectors, 6, 34-34 (2013-02-13)
Current development efforts of subunit vaccines against Toxoplasma gondii, the etiological agent of toxoplasmosis, have been focused mainly on tachyzoite surface antigen 1 (SAG1). Since microparticles made from poly (lactide-co-glycolide) (PLG) polymers have been developed as safe, potent adjuvants or
Vanna Sanna et al.
International journal of nanomedicine, 7, 5501-5516 (2012-10-25)
Resveratrol, like other natural polyphenols, is an extremely photosensitive compound with low chemical stability, which limits the therapeutic application of its beneficial effects. The development of innovative formulation strategies, able to overcome physicochemical and pharmacokinetic limitations of this compound, may
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service