NA2020
GenElute™ Blood Genomic DNA Kit
sufficient for 350 purifications
Synonym(s):
Blood Genomic DNA, Gen Elute
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
UNSPSC Code:
41105501
NACRES:
NA.52
Recommended Products
usage
sufficient for 350 purifications
Quality Level
technique(s)
DNA purification: suitable
storage temp.
15-25°C
room temp
Looking for similar products? Visit Product Comparison Guide
General description
The GenElute Blood Genomic DNA kit provides a simple and convenient way to isolate pure genomic DNA from fresh or aged (older than 24 hours) whole blood. The kit combines the advantages of silica binding with a microspin format, and eliminates the need for expensive resins, alcohol precipitation, and hazardous organic compounds such as phenol and chloroform.
Application
The purified genomic DNA is ready for downstream applications such as:
- restriction endonuclease digestions
- PCR
- Southern blots
- sequencing reactions
- cloning
Features and Benefits
- Starting material: Up to 200 μl of fresh or aged blood
- Expected yield: Up to 10 μg
- Elution volume: 400 μl
- Time required: <40 min
- A260/A280 ratio: 1.6 - 1.9
- Compatible with many anticoagulants, including EDTA, Heparin, and Sodium Citrate
- Typical yields 4-10 μg from 200 μL of whole blood
- Pure genomic DNA with a A260/A280 ratio between 1.6 and 1.9
- Isolate DNA up to 50 kb in length
Principle
The starting material is lysed in a chaotropic salt-containing solution to ensure the thorough denaturation of macromolecules. The addition of ethanol causes the DNA to bind when the lysate is spun through a silica membrane in a microcentrifuge tube. A Prewash Solution is provided to help remove contaminants that are associated with aged (older than 24 hours) whole blood samples. After washing to remove contaminants, the DNA is eluted in 200 mL of a Tris-EDTA solution.
The expected yields of genomic DNA will vary depending on the amount and nature of the starting material used (for example, 4 to 10 μg of RNase A-treated DNA can be isolated from 200 μl of fresh whole blood in less than one hour). DNA purified with this kit has an A260/A280 ratio between 1.6 and 1.9 and can be up to 50 kb in length.
The expected yields of genomic DNA will vary depending on the amount and nature of the starting material used (for example, 4 to 10 μg of RNase A-treated DNA can be isolated from 200 μl of fresh whole blood in less than one hour). DNA purified with this kit has an A260/A280 ratio between 1.6 and 1.9 and can be up to 50 kb in length.
Other Notes
For additional information, please see www.sigma-aldrich.com/genomicdna.
Legal Information
GenElute is a trademark of Sigma-Aldrich Co. LLC
recommended
related product
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Miri Cohen-Zinder et al.
Physiological genomics, 48(5), 367-376 (2016-03-20)
Ecological and economic concerns drive the need to improve feed utilization by domestic animals. Residual feed intake (RFI) is one of the most acceptable measures for feed efficiency (FE). However, phenotyping RFI-related traits is complex and expensive and requires special
S K Sahu et al.
Oncogenesis, 5, e225-e225 (2016-05-10)
Tumor suppressor p53 is a critical player in the fight against cancer as it controls the cell cycle check point, apoptotic pathways and genomic stability. It is known to be the most frequently mutated gene in a wide variety of
Disha Bhange et al.
Frontiers in microbiology, 12, 779472-779472 (2021-12-14)
In a multicentric, observational, investigator-blinded, and longitudinal clinical study of 764 ART-naïve subjects, we identified nine different promoter variant strains of HIV-1 subtype C (HIV-1C) emerging in the Indian population, with some of these variants being reported for the first
C Castellini et al.
Theriogenology, 78(4), 715-723 (2012-06-19)
There is increasing evidence that the particulate fraction of seminal plasma plays an important role in reproduction of several mammalian species. However, the origin and role of these granules in the physiology of rabbit spermatozoa is partially unknown. The aim
Anika Daing et al.
Journal of periodontal & implant science, 42(5), 151-157 (2012-11-28)
Cyclooxygenase (COX) enzyme catalyzes the production of prostaglandins, which are important mediators of tissue destruction in periodontitis. Single nucleotide polymorphisms of COX2 enzyme have been associated with increasing susceptibility to inflammatory diseases. The present study evaluates the association of two
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service