Skip to Content
Merck
All Photos(3)

Key Documents

N9003

Sigma-Aldrich

4-Nitrophenyl N-acetyl-β-D-galactosaminide

≥98%

Synonym(s):

4-Nitrophenyl 2-acetamido-2-deoxy-β-D-galactopyranoside, p-Nitrophenyl N-acetyl-β-D-galactosaminide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N2O8
CAS Number:
Molecular Weight:
342.30
Beilstein:
96194
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.28

Quality Level

Assay

≥98%

form

powder

solubility

DMF: 50 mg/mL

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H]1[C@@H](O)[C@@H](O)[C@@H](CO)O[C@H]1Oc2ccc(cc2)[N+]([O-])=O

InChI

1S/C14H18N2O8/c1-7(18)15-11-13(20)12(19)10(6-17)24-14(11)23-9-4-2-8(3-5-9)16(21)22/h2-5,10-14,17,19-20H,6H2,1H3,(H,15,18)/t10-,11-,12+,13-,14-/m1/s1

InChI key

OMRLTNCLYHKQCK-RKQHYHRCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Nitrophenyl N-acetyl-β-D-galactosaminide is a chromogenic substrate for enzymes N-acetyl-β-D-galactosaminidase and N-Acetyl-β-D-hexosaminidase. It is membrane-permeable and serves as a substrate for the enzyme 6-sulfotransferase in Golgi vesicles.

Application

4-Nitrophenyl N-acetyl-β-D-galactosaminide has been used:
  • as a synthetic substrate in the glycosyl hydrolase assay of Yersinia pseudotuberculosis protein
  • as a substrate in -β-hexosaminidase assay in adenocarcinomic human alveolar basal epithelial cells A549
  • as a substrate for N-acetyl-β-D-galactosaminidase assay in rat kidney tissue extracts

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lysosomal enzymes are decreased in the kidney of diabetic rats
Peres GB, et al.
Biochimica et Biophysica Acta (BBA)-Molecular Basis of Disease, 1832(1), 85-95 (2013)
Loss of a biofilm-inhibiting glycosyl hydrolase during the emergence of Yersinia pestis
Erickson DL, et al.
Journal of Bacteriology, 190(24), 8163-8170 (2008)
Heterologous expression and characterization of an N-acetyl-beta-D-hexosaminidase from Lactococcus lactis ssp. lactis IL1403
Nguyen HA, et al.
Journal of Agricultural and Food Chemistry, 60(12), 3275-3281 (2012)
R D Roer et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 128(4), 683-690 (2001-04-06)
We have previously demonstrated a marked change in sugar moieties of glycoproteins of the cuticle of the blue crab, Callinectes sapidus, between 0.5 and 3 h post-ecdysis. The present study has identified a glycosidase that appears in the cuticle during
S K Chatterjee et al.
Cancer, 49(1), 128-135 (1982-01-01)
Levels and isoenzyme profiles of beta-hexosaminidase were compared in extracts from normal ovarian and ovarian epithelial tumors. The specific activities of beta-hexosaminidase were significantly (P less than 0.001) higher in malignant than in normal ovarian tissues. The enzyme levels of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service