Skip to Content
Merck
All Photos(6)

Key Documents

N1252

Sigma-Aldrich

4-Nitrophenyl β-D-galacto­pyran­oside

≥98% (enzymatic),≥98% (TLC), powder

Synonym(s):

p-Nitrophenyl β-D-galacto­pyran­oside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO8
CAS Number:
Molecular Weight:
301.25
Beilstein:
92213
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

4-Nitrophenyl β-D-galacto­pyran­oside, ≥98% (enzymatic)

Assay

≥98% (TLC)
≥98% (enzymatic)

form

powder

solubility

water: 10 mg/mL, clear, colorless to very faintly green

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2ccc(cc2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C12H15NO8/c14-5-8-9(15)10(16)11(17)12(21-8)20-7-3-1-6(2-4-7)13(18)19/h1-4,8-12,14-17H,5H2/t8-,9+,10+,11-,12-/m1/s1

InChI key

IFBHRQDFSNCLOZ-YBXAARCKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Nitrophenyl β-D-galactopyranoside has been used:
  • as a substrate to assess the activity of glycosaminoglycan (GAG)-degrading enzymes
  • as a substrate to study the kinetic properties of recombinant Leuconostoc mesenteroides glycosidase (BgLm1) and determine β-glucosidase activity
  • to prepare substrate solution in a modified universal buffer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification, purification and characterization of a novel glycosidase (BgLm1) from Leuconostoc mesenteroides
del Pino-Garcia R, et al.
LWT--Food Science and Technology null
A high-throughput microplate assay for simultaneous colorimetric quantification of multiple enzyme activities in soil
Popova IR and Deng S
Applied soil ecology : a section of Agriculture, Ecosystems & Environment null
Radosław Kowalewski et al.
Journal of vascular research, 43(1), 95-100 (2005-11-19)
The abdominal aortic aneurysm (AAA) wall represents an extreme example of arterial remodeling with disturbed elastin, collagen and proteoglycan metabolism. The aim of this study was to evaluate enzymes involved in the degradation of glycosaminoglycan chains and core proteins of
Daniel M Stoebel et al.
Genetics, 178(3), 1653-1660 (2008-02-05)
Transcriptional regulatory networks allow bacteria to express proteins only when they are needed. Adaptive hypotheses explaining the evolution of regulatory networks assume that unneeded expression is costly and therefore decreases fitness, but the proximate cause of this cost is not
Lan Guan et al.
Biochemistry, 42(6), 1377-1382 (2003-02-13)
Major determinants for substrate recognition by the lactose permease of Escherichia coli are at the interface between helices IV (Glu126, Ala122), V (Arg144, Cys148), and VIII (Glu269). We demonstrate here that Trp151, one turn of helix V removed from Cys148

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service