Skip to Content
Merck
All Photos(4)

Key Documents

N6538

Sigma-Aldrich

Anti-NR4A2 antibody produced in rabbit

enhanced validation

~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-HZF-3, Anti-NOT, Anti-NURR1, Anti-RNR1, Anti-TINUR

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~70 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.5 mg/mL

technique(s)

indirect immunofluorescence: 4-8 μg/mL using HEK-293T cell lysate expressing human NR4A2
western blot: 1-2 μg/mL using HEK-293T cell lysate expressing human NR4A2

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR4A2(4929)

General description

NR4A2 is encoded by the gene mapped to human chromosome 2q24.1.
Nuclear receptor subfamily 4 group A member 2 (NR4A2) is part of transcriptional regulator subfamily, under the family of nuclear hormone receptors. It is a transcription factor expressed in the brain and T-cells.

Specificity

Anti-NR4A2 specifically recognizes human NR4A2.

Application

Anti-NR4A2 antibody produced in rabbit has also been used in immunohistochemistry (IHC).
Anti-NR4A2 antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Nuclear receptor subfamily 4 group A member 2 (NR4A2) acts as an activator of tyrosine hydroxylase. It is crucial for the process of DNA repair by melanocortin-1 receptor.
Nuclear receptor subfamily 4 group A member 2 (NR4A2) receptors function as transcription factors. NR4A2 binds the NGFI-B (nerve growth factor-induced clone B) response element (NBRE) sequence (AAAGGTCA) in the form of monomer. NR4A2 is induced by multiple extracellular signals, including fatty acids, stress, prostaglandins, growth factors, calcium, inflammatory cytokines, peptide hormones, and neurotransmitters in a cell type specific manner. Cyclic adenosine monophosphate (cAMP) triggers the hepatic expression of NR4A2 in response to glucagon and fasting. Diabetic mice with elevated gluconeogenesis also show high levels of NR4A2. NR4A2 is essential for the differentiation of nigral dopaminergic neurons. Mice lacking Nurr1 failed to generate midbrain dopaminergic neurons. NR4A2 mutations have been associated with disorders related to dopaminergic dysfunction, including Parkinson′s disease, schizophrenia and manic depression.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. Storage in “frost-free” freezers is also not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The NR4A subgroup: immediate early response genes with pleiotropic physiological roles
Maxwell MA and Muscat GEO
Nuclear Receptor Signaling, 4(1), nrs-04002 (2006)
Extended mutation analysis and association studies of Nurr1 (NR4A2) in Parkinson disease
Hering R, et al.
Neurology, 62(7), 1231-1232 (2004)
M Lappas
Placenta, 35(11), 866-875 (2014-09-10)
Members of the NR4A subfamily are involved in a wide range of diseases including obesity and diabetes. The aim of this study was to determine the effect of maternal obesity and gestational diabetes mellitus (GDM) on the expression of the
Acid ceramidase (ASAH1) is a global regulator of steroidogenic capacity and adrenocortical gene expression
Lucki NC, et al.
Molecular Endocrinology, 26(2), 228-243 (2012)
Kasturee Jagirdar et al.
PloS one, 8(11), e78075-e78075 (2013-11-14)
Ultraviolet radiation (UVR) is one of the most common mutagens encountered by humans and induces the formation of cyclobutane pyrimidine dimers (CPDs) and pyrimidine-(6-4)-pyrimidone photoproduct (6-4PP) lesions in the genomic DNA. To prevent the accumulation of deleterious mutations these lesions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service