Skip to Content
Merck
All Photos(1)

Documents

47435

Sigma-Aldrich

Fmoc-β-(3-pyridyl)-D-Ala-OH

purum, ≥98.0% (HPLC)

Synonym(s):

Fmoc-3-(3-pyridyl)-D-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H20N2O4
CAS Number:
Molecular Weight:
388.42
Beilstein:
9223259
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

grade

purum

Quality Level

Assay

≥98.0% (HPLC)

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@@H](Cc1cccnc1)NC(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C23H20N2O4/c26-22(27)21(12-15-6-5-11-24-13-15)25-23(28)29-14-20-18-9-3-1-7-16(18)17-8-2-4-10-19(17)20/h1-11,13,20-21H,12,14H2,(H,25,28)(H,26,27)/t21-/m1/s1

InChI key

JQLPMTXRCLXOJO-OAQYLSRUSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service