Skip to Content
Merck
All Photos(4)

Key Documents

N3877

Sigma-Aldrich

Potassium 4-nitrophenyl sulfate

sulfatase substrate, chromogenic, ≥98% (TLC), powder

Synonym(s):

4-Nitrophenyl sulfate potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NO2C6H4OSO2OK
CAS Number:
Molecular Weight:
257.26
Beilstein:
3786392
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

Potassium 4-nitrophenyl sulfate, sulfatase substrate

Assay

≥98% (TLC)

form

powder

mp

246-250 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless to very faintly greenish-yellow (to Very Light Yellow)

shipped in

wet ice

storage temp.

−20°C

SMILES string

[K+].[O-][N+](=O)c1ccc(OS([O-])(=O)=O)cc1

InChI

1S/C6H5NO6S.K/c8-7(9)5-1-3-6(4-2-5)13-14(10,11)12;/h1-4H,(H,10,11,12);/q;+1/p-1

InChI key

BITVAZYUWRLLCN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Potassium 4-nitrophenyl sulfate has been used:
  • as a substrate to measure arylsulfatase activity in cell-free coelomic fluid
  • as a substrate for p-nitrophenyl glycoside-based enzyme assay
  • to inhibit arylsulfatase
  • as an inorganic analog of organic aryl ester sulphate

Substrates

Chromogenic sulfatase substrate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J O Falkinham
International journal of systematic bacteriology, 40(1), 66-70 (1990-01-01)
A rapid (3-h) arylsulfatase assay for cell suspensions of mycobacteria, in which p-nitrophenyl sulfate is used as the substrate, was developed. Arylsulfatase activity was found in cell suspensions of representative strains of Mycobacterium avium, Mycobacterium intracellulare, and Mycobacterium scrofulaceum grown
N Sakuma-Sawada et al.
Research communications in molecular pathology and pharmacology, 97(2), 131-138 (1997-08-01)
Hepatic uptake of the sulfate conjugate of p-nitrophenol (p-NPsul) has been studied. Uptake clearance of p-NPsul by isolated rat hepatocytes was dependent on p-NPsul concentration, suggesting carrier-mediated transport. The uptake of p-NPsul by isolated rat hepatocytes was inhibited by acetaminophen
M P Kung et al.
Endocrinology, 122(4), 1195-1200 (1988-04-01)
Subcellular preparations from rat liver, brain, and kidney and from human liver were tested for their ability to desulfate T3 sulfate (T3SO4). Activity was found associated with the microsomal fraction: rat liver was the most active, hydrolyzing 76 pmol/min.mg protein
H X Zhang et al.
Drug metabolism and disposition: the biological fate of chemicals, 19(2), 473-477 (1991-03-01)
Although numerous previous reports have characterized the mammalian biotransformation of the organophosphorus insecticides parathion and methyl parathion, questions still remain regarding the toxicological significance of certain metabolic pathways in vivo. The present study utilized rat liver perfusions in order to
Shina Caroline Lynn Kamerlin
The Journal of organic chemistry, 76(22), 9228-9238 (2011-10-11)
Both phosphoryl and sulfuryl transfers are ubiquitous in biology, being involved in a wide range of processes, ranging from cell division to apoptosis. Additionally, it is becoming increasingly clear that enzymes that can catalyze phosphoryl transfer can often cross-catalyze sulfuryl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service