Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
4-Nitrophenyl β-D-glucuronide
Synonym(s):
4-Nitrophenyl-β-D-glucopyranosiduronic acid, PNPG
Empirical Formula (Hill Notation):
C
12
H
13
NO
9
CAS Number:
10344-94-2
Molecular Weight:
315.23
EC Number:
233-753-0
Products
Product Category
substrates (2)
Brand
Sigma-Aldrich (2)
Formula Weight
Physical Form
Purity
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
N1627
4-Nitrophenyl β-D-glucuronide,
≥98% (TLC)
Expand
View Pricing
73677
4-Nitrophenyl β-D-glucuronide,
β-glucuronidase substrate, chromogenic, ≥99.0% (TLC), powder or crystals, suitable as substrate for β-glucuronidase test
Expand
View Pricing