Skip to Content
Merck
All Photos(1)

Key Documents

N0290

Sigma-Aldrich

Nitazoxanide

≥98% (HPLC)

Synonym(s):

NTZ; 2-(Acetyloxy)-N-(5-nitro-2-thiazolyl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9N3O5S
CAS Number:
Molecular Weight:
307.28
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

originator

Romark

storage temp.

2-8°C

SMILES string

CC(=O)Oc1ccccc1C(=O)Nc2ncc(s2)[N+]([O-])=O

InChI

1S/C12H9N3O5S/c1-7(16)20-9-5-3-2-4-8(9)11(17)14-12-13-6-10(21-12)15(18)19/h2-6H,1H3,(H,13,14,17)

InChI key

YQNQNVDNTFHQSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nitazoxanide (NTZ), a thiazolide compound is a antiparasitic drug with structure similar to niclosamide.

Application

Nitazoxanide has been used:
  • to test its anti-viral activity against chikungunya virus
  • as an antiprotozoal agent to test its effect on cell viability in various cancer cell lines
  • to test its effect on human cytomegalovirus (HCMV) infected human fibroblast HFF cells

Biochem/physiol Actions

Nitazoxanide (NTZ) promotes autophagy by acting on kinase based signaling pathways and acts on mammalian target of rapamycin complex 1 (mTORC1) in Mycobacteria. It has anti-viral property and effectively halts entry and release of chikungunya virus in in vitro studies. NTZ also inhibits Japanese encephalitis virus (JEV) infection in early stages and has the potential to treat other viral infections including dengue, hepatitis B (HBV), coronavirus and human immunodeficiency virus (HIV). It has antineoplastic functionality and may induce apoptosis by promoting proto-oncogene c-Myc inhibition resulting in tumor suppression.
Nitazoxanide is an inhibitor of pyruvate-ferredoxin oxidoreductase (PFOR); FDA approved anti-parasitic drug (2002). Recent work (C & EN Sept. 14, 2009, p. 28) highlights that NTZ kills non-replicating and replicating TB bacteria and no apparent resistance is detected.

Features and Benefits

This compound was developed by Romark. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Patrick Basu et al.
The American journal of gastroenterology, 106(11), 1970-1975 (2011-10-13)
Resistance to standard Helicobacter pylori (HP) treatment regimens has led to unsatisfactory cure rates in HP-infected patients. This study was designed to evaluate a novel four-drug regimen (three antibiotics and a proton pump inhibitor (PPI)) for eradication of HP infection
Irit Krause et al.
The Pediatric infectious disease journal, 31(11), 1135-1138 (2012-07-20)
Cryptosporidium parvum is a common cause of diarrhea. In immunocompetent individuals, spontaneous recovery is the rule. In immunocompromised patients, it may cause a serious disease. Data on cryptosporidiosis in children after solid organ transplantation are few. We report on 6
Nitazoxanide stimulates autophagy and inhibits mTORC1 signaling and intracellular proliferation of Mycobacterium tuberculosis
Lam KYK
PLoS Pathogens, 8(5), 340-351 (2012)
Vanessa R Anderson et al.
Drugs, 67(13), 1947-1967 (2007-08-29)
Nitazoxanide (Alinia, Daxon, Dexidex, Paramix, Kidonax, Colufase, Annita) has in vitro activity against a variety of microorganisms, including a broad range of protozoa and helminths. Nitazoxanide is effective in the treatment of protozoal and helminthic infections, including Cryptosporidium parvum or
Yijun Zhu et al.
Cell host & microbe, 31(1), 18-32 (2022-12-23)
Recent studies show gut microbiota-dependent metabolism of dietary phenylalanine into phenylacetic acid (PAA) is critical in phenylacetylglutamine (PAGln) production, a metabolite linked to atherosclerotic cardiovascular disease (ASCVD). Accordingly, microbial enzymes involved in this transformation are of interest. Using genetic manipulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service