Skip to Content
Merck
All Photos(1)

Key Documents

D198501

Sigma-Aldrich

2,4-Dinitrophenol

moistened with water, ≥98.0%

Synonym(s):

α-Dinitrophenol, 2,4-DNP, DNP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3OH
CAS Number:
Molecular Weight:
184.11
Beilstein:
1246142
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.35 (vs air)

Assay

≥98.0%

contains

≥15% water

mp

108-112 °C (lit.)

SMILES string

Oc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H4N2O5/c9-6-2-1-4(7(10)11)3-5(6)8(12)13/h1-3,9H

InChI key

UFBJCMHMOXMLKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4-Dinitrophenol (DNP) can be used:
  • As a reactant for catalytic reduction reactions.
  • To activate carboxylic acids by converting them into dinitrophenyl (DNP) esters.
  • To prepare the corresponding ester via acylation reaction using isobutyric anhydride catalyzed by hafnium triflate.
  • As an effective cocatalyst to accelerate the activity and enantioselectivity of primary amine organocatalyst derived from natural primary amino acids for direct asymmetric aldol reaction.
  • As an alternative activator to tetrazoles in the reaction of phosphoroamidites with nucleosides.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Acute 1 - Desen. Expl. 4 - STOT RE 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chemical-hydrothermal synthesis of oval-shaped graphene/ZnO quantum hybrids and their photocatalytic performances.
Van T D, et al.
Catalysis Communications, 101, 102-106 (2017)
Synthesis and stereochemical assignment of geraniol-and nerol-derived Cygerol enantiomers.
Sukhanova A A, et al.
Tetrahedron Asymmetry, 28(12), 1834-1841 (2017)
Hafnium inspired activation of highly hindered anhydrides in the acylation of alcohols and polyols.
Mensah E, et al.
Tetrahedron Letters, 59(12), 1094-1098 (2018)
2, 4-Dinitrophenol: a novel activating reagent in nucleotide synthesis via the phosphoramidite route. Design of new effective phosphitylating reagents.
Dabkowski W, et al.
Tetrahedron Letters, 41(39), 7535-7539 (2000)
Photo-Enhanced Selective Reduction of Nitroarenes Over Pt/ZnO Catalyst.
Van T D, et al.
Catalysis Letters, 147(9), 2440-2447 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service