Skip to Content
Merck
All Photos(1)

Key Documents

TR0100

Sigma-Aldrich

Serum Triglyceride Determination Kit

1 kit sufficient for 250 tests

Synonym(s):

Triglyceride and Free Glycerol Kits and Reagents

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

usage

1 kit sufficient for 250 tests

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

The Serum Triglyceride Determination Kit is for the quantitative enzymatic measurement of glycerol, true triglycerides, and total triglycerides in serum or plasma at 540 nm. Triglycerides, esters of fatty acids and glycerol,1 do not circulate freely in plasma, but are bound to proteins and transported as macromolecular complexes called lipoproteins.2 Methods for triglyceride determination generally involve enzymatic3 or alkaline4 hydrolysis of triglycerides to glycerol and free fatty acids, followed by either chemical or enzymatic measurement of the glycerol released.

Application

A Serum Triglyceride Determination Kit was used to determine the level of triglycerides in whole-body homogenates of Drosophila melanogaster.
The Serum Triglyceride Determination Kit can be used for the measurement of glycerol, true triglycerides, or total triglycerides in serum or plasma. The procedure involves enzymatic hydrolysis by lipase of the triglycerides to glycerol and free fatty acids. The glycerol produced is then measured by coupled enzyme reactions. Many of the triglyceride reagents which are commercially available, do not differentiate between endogenous glycerol and glycerol derived by hydrolytic action of lipase on glycerides.
Triglycerides are first hydrolyzed by lipoprotein lipase to glycerol and free fatty acids. Glycerol is then phosphorylated by adenosine-5′-triphosphate (ATP) forming glycerol-1-phosphate (G-1-P) and adenosine-5′-diphosphate (ADP) in the reaction catalyzed by glycerol kinase (GK). G-1-P is then oxidized by glycerol phosphate oxidase (GPO) to dihydroxy-acetone phosphate (DAP) and hydrogen peroxide (H2O2). Peroxidase (POD) catalyzes the coupling of H2O2 with 4-aminoantipyrine (4-AAP) and sodium
N-ethyl-N-(3-sulfopropyl) m-anisidine (ESPA) to produce a quinoneimine dye that shows an absorbance maximum at 540 nm. The increase in absorbance at 540 nm is directly proportional to triglyceride concentration of the sample.
The triglyceride and free glycerol kits are for the quantitative determination of glycerol, total triglycerides or free triglycerides.

Packaging

The kit also includes sufficient reagent for an additional 250 free glyceride tests for true triglyceride determination.

Suitability

Suitable for the quantitative determination of glycerol, total triglycerides or free triglycerides in serum or plasma

Principle

The Serum Triglyceride Determination Kit can be used for the measurement of glycerol, true triglycerides, or total triglycerides in serum or plasma. The procedure involves enzymatic hydrolysis by lipase of the triglycerides to glycerol and free fatty acids. The glycerol produced is then measured by coupled enzyme reactions. Many of the triglyceride reagents which are commercially available, do not differentiate between endogenous glycerol and glycerol derived by hydrolytic action of lipase on glycerides. Triglycerides are first hydrolyzed by lipoprotein lipase to glycerol and free fatty acids. Glycerol is then phosphorylated by adenosine-5′-triphosphate (ATP) forming glycerol-1-phosphate (G-1-P) and adenosine-5′-diphosphate (ADP) in the reaction catalyzed by glycerol kinase (GK). G-1-P is then oxidized by glycerol phosphate oxidase (GPO) to dihydroxy-acetone phosphate (DAP) and hydrogen peroxide (H2O2). Peroxidase (POD) catalyzes the coupling of H2O2 with 4-aminoantipyrine (4-AAP) and sodium N-ethyl-N-(3-sulfopropyl) m-anisidine (ESPA) to produce a quinoneimine dye that shows an absorbance maximum at 540 nm. The increase in absorbance at 540 nm is directly proportional to triglyceride concentration of the sample.

Linkage

In addition to kits, the individual reagents and glycerol standard are available separately when fewer reactions are needed.

Kit Components Also Available Separately

Product No.
Description
SDS

  • F6428Free Glycerol Reagent, used for quantitative enzymatic determination of glycerol 10 x 40SDS

  • T2449Triglyceride Reagent, used for quantitative enzymatic determination of triglyerides 5 x 10SDS

standard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dorothy A Kieffer et al.
The Journal of nutrition, 146(12), 2445-2460 (2016-11-01)
Enzyme-treated wheat bran (ETWB) contains a fermentable dietary fiber previously shown to decrease liver triglycerides (TGs) and modify the gut microbiome in mice. It is not clear which mechanisms explain how ETWB feeding affects hepatic metabolism, but factors (i.e., xenometabolites)
Edith Grosbellet et al.
Chronobiology international, 32(5), 637-649 (2015-06-03)
Leptin may affect central and/or peripheral timing, in addition to its well-known regulatory effects on metabolism. Here, we investigated whether leptin can impact rhythmicity of blood glucose and lipids. For that purpose, daily variations of blood glucose and lipids were
Colin S McCoin et al.
American journal of physiology. Endocrinology and metabolism, 317(2), E298-E311 (2019-05-01)
The impact of sexual dimorphism and mitophagy on hepatic mitochondrial adaptations during the treatment of steatosis with physical activity are largely unknown. Here, we tested if deficiencies in liver-specific peroxisome proliferative activated-receptor-γ coactivator-1α (PGC-1α), a transcriptional coactivator of biogenesis, and
Sarah I Jacob et al.
Journal of veterinary internal medicine, 32(3), 1215-1233 (2018-03-25)
Metabolomics, the study of small-molecule metabolites, has increased understanding of human metabolic diseases, but has not been used to study equine metabolic syndrome (EMS). (1) To examine the serum metabolome of Welsh Ponies with and without insulin dysregulation before and
Frank K Huynh et al.
Journal of inherited metabolic disease, 41(1), 59-72 (2017-07-21)
Several inherited metabolic disorders are associated with an accumulation of reactive acyl-CoA metabolites that can non-enzymatically react with lysine residues to modify proteins. While the role of acetylation is well-studied, the pathophysiological relevance of more recently discovered acyl modifications, including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service