Skip to Content
Merck
All Photos(1)

Key Documents

M9004

Sigma-Aldrich

Malic Dehydrogenase from bovine heart

ammonium sulfate suspension, 2000-4000 units/mg protein (modified Warburg-Christian)

Synonym(s):

L-Malate:NAD+ oxidoreductase, MDH

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bovine heart

Quality Level

form

ammonium sulfate suspension

specific activity

2000-4000 units/mg protein (modified Warburg-Christian)

mol wt

35 kDa

impurities

≤0.01% Glutamic-Oxalacetic Transaminase
≤0.01% Glutamic-Pyruvic Transaminase

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Malic Dehydrogenase from bovine heart contains a histidine residue at the NAD-binding active site which is critical for activity. When this histidine is mutated a loss in activity is observed.
Malic dehydrogenase is a cytoplasmic isozyme and an important catalyst in the tricarboxylic acid cycle.

Application

Malic dehydrogenase has been used in a study to assess a flow injection system for on-line monitoring of fumaric acid in biological processes. It has also been used in a study to investigate a root-knot nematode parasitizing peanut in Texas.

Unit Definition

One unit will convert 1.0 μmole of oxalacetate and β-NADH to L-malate and β-NAD per min at pH 7.5 at 25 °C.

Physical form

Suspension in 3 M (NH4)2SO4 - 0.01 M KH2PO4 solution, pH 7.3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eisaku Hokazono et al.
Clinica chimica acta; international journal of clinical chemistry, 412(15-16), 1436-1440 (2011-05-10)
In human serum, as for phospholipids not containing choline, phosphatidylethanolamine (PE) exists approximately 5% in a whole phospholipid. PE is well known as one of the main components of biological membranes, and also plays important roles that contribute to apoptosis
J D Eisenback et al.
Journal of nematology, 35(4), 395-403 (2003-12-01)
Meloidogyne haplanaria n. sp. is described and illustrated from specimens parasitizing peanut in Texas. The perineal pattern of the female is rounded to oval with a dorsal arch that is high and rounded except for striae near the vulva, which
Isoelectric points and molecular weights of proteins: a new table.
Righetti, P.G., and Tudor, G.
Journal of Chromatography A, 220, 115-194 (1981)
Z Y Chen et al.
Plant physiology, 112(2), 677-684 (1996-10-01)
At low-CO2 (air) conditions, the unicellular green alga Chlamydomonas reinhardtii acquires the ability to raise its internal inorganic carbon concentration. To study this adaptation to low CO2, cDNA clones induced under low-CO2 growth conditions were selected through differential screening. One
Flow injection system for on-line monitoring of fumaric acid in biological processes
Rhee, J.
Analytica Chimica Acta, 499, 71-80 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service