Skip to Content
Merck
All Photos(5)

Documents

M4263

Sigma-Aldrich

Malonyl coenzyme A lithium salt

≥90% (HPLC)

Synonym(s):

Malonyl CoA lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H38N7O19P3S · xLi+
CAS Number:
Molecular Weight:
853.58 (free acid basis)
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥90% (HPLC)

solubility

H2O: soluble 50 mg/mL protein, clear, colorless

storage temp.

−20°C

SMILES string

[Li].CC(C)(COP(O)(=O)OP(O)(=O)OCC1OC(C(O)C1OP(O)(O)=O)n2cnc3c(N)ncnc23)C(O)C(=O)NCCC(=O)NCCSC(=O)CC(O)=O

InChI

1S/C24H38N7O19P3S.Li/c1-24(2,19(37)22(38)27-4-3-13(32)26-5-6-54-15(35)7-14(33)34)9-47-53(44,45)50-52(42,43)46-8-12-18(49-51(39,40)41)17(36)23(48-12)31-11-30-16-20(25)28-10-29-21(16)31;/h10-12,17-19,23,36-37H,3-9H2,1-2H3,(H,26,32)(H,27,38)(H,33,34)(H,42,43)(H,44,45)(H2,25,28,29)(H2,39,40,41);

InChI key

OPIJLICRFQMMJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Malonyl coenzyme A lithium salt has been used:
  • in Krebs Ringer bicarbonate medium for preincubation of trypsinized and re-suspended fibroblast for fatty acid oxidation assay
  • in HEPES (4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid) buffer used for scintillation proximity assay for fatty acid synthase
  • as an internal standard in the reaction mixture used for succinyl-CoA ligase assay

Biochem/physiol Actions

Coenzyme A functions as an acyl group carrier, acetyl-CoA. Malonyl Coenzyme A is a coenzyme A derivative that is utilized in fatty acid and polyketide synthesis and in the transport of α-ketoglutarate across the mitochondrial membrane. Malonyl CoA is formed by the Acetyl CoA Carboxylase-mediated carboxylation of acetyl CoA. Malonyl-CoA is exclusively used as the extender unit in the synthesis of bacterial aromatic polyketides.
Malonyl coenzyme A is a coenzyme A derivative that is utilized in fatty acid and polyketide synthesis and in the transport of α-ketoglutarate across the mitochondrial membrane. Malonyl CoA is formed by the Acetyl CoA Carboxylase-mediated carboxylation of acetyl CoA. Metabolism of glucose also yields malonyl coenzyme A. It allosterically blocks the action of carnitine palmitoyltransferase 1, which affects the transfer of long chain fatty acids into mitochondria. Inactivation of fatty acid synthase results in excess malonyl coenzyme A which leads to anorexia.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R G Summers et al.
Biochemistry, 34(29), 9389-9402 (1995-07-25)
Streptomyces glaucescens, a Gram-positive soil bacterium, produces the polyketide antibiotic tetracenomycin (Tcm) C. To study possible biochemical connections between the biosynthesis of bacterial fatty acids and polyketides, the abundant acyl carrier protein (ACP) detected throughout the growth of the tetracenomycin
J Kalervo Hiltunen et al.
Biochimica et biophysica acta, 1797(6-7), 1195-1202 (2010-03-17)
Recent studies have revealed that mitochondria are able to synthesize fatty acids in a malonyl-CoA/acyl carrier protein (ACP)-dependent manner. This pathway resembles bacterial fatty acid synthesis (FAS) type II, which uses discrete, nuclearly encoded proteins. Experimental evidence, obtained mainly through
Characterization of fatty acid synthase activity using scintillation proximity
Weiss DR and Glickman JF
Assay and Drug Development Technologies, 1(1-2), 161-166 (2003)
Succinyl-CoA synthetase (SUCLA2) deficiency in two siblings with impaired activity of other mitochondrial oxidative enzymes in skeletal muscle without mitochondrial DNA depletion
Huang X, et al.
Molecular Genetics and Metabolism, 120(3), 213-222 (2017)
Gary D Lopaschuk et al.
Pharmacological reviews, 62(2), 237-264 (2010-04-16)
The central nervous system mediates energy balance (energy intake and energy expenditure) in the body; the hypothalamus has a key role in this process. Recent evidence has demonstrated an important role for hypothalamic malonyl CoA in mediating energy balance. Malonyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service